Skip to Content
Merck
All Photos(1)

Documents

B6917

Sigma-Aldrich

Bovine Serum Albumin

lyophilized powder, suitable for (for molecular biology), Non-acetylated

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

bovine

grade

for molecular biology

Assay

≥98% (agarose gel electrophoresis)

form

lyophilized powder

quality

deacetylated

mol wt

~66 kDa

purified by

heat shock fractionation

packaging

glass bottle of

origin

USA origin

technique(s)

electrophoresis: suitable-98% using Agarose

impurities

enzymatic impurity, none detected

pH

7

solubility

water: soluble (50 mg/ml)

suitability

suitable for (for molecular biology)

UniProt accession no.

foreign activity

Alkaline Phosphatase free
Alkaline phosphatase and peroxidase, none detected
DNase free
DNase, RNase, and protease, none detected
Peroxidase free
Protease free
RNase free

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Bovine Serum Albumin (BSA) is a plasma protein produced in the bovine liver that has a globular, non-glycosylated α-helical structure. It consists of a single polypeptide chain with 583 amino acid residues and 17 cysteine residues that form the cross-linked disulfide bridges, stabilizing its structure. The BSA protein comprises three structurally distinct domains (I, II, and III), each of which contains two sub-domains (A and B) arranged in a heart-shaped architecture.

Application

Bovine Serum Albumin (BSA) has been used
  • as a component of phosphate buffer saline (PBS) solution for dissolving tumor necrosis factor α (TNFα), which was added as a supplement for culture media
  • as a blocking agent to reduce non-specific binding during the albumin sensor assay
  • as a blocking agent in the immunofluorescence (IF) assay
  • as a protein standard in the Bradford assay

Biochem/physiol Actions

Bovine Serum Albumin (BSA) is a versatile protein commonly utilized in cell and tissue culture media as a supplement. It helps in lipid solubilization and stabilizing other proteins. BSA is also a carrier for small molecules such as steroids, fatty acids, and thyroid hormones. It serves as a standard protein for immunochemical tests to determine protein concentration in a solution and calibration studies.
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Features and Benefits

This BSA product for molecular biology is tested for the following properties:
* Alkaline Phosphatase-free
* DNase-free / Exonuclease-free
* Nickase-free / Endonuclease-free
* Peroxidase-free
* Protease-free
* RNase-free

Preparation Note

Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fadi Jacob et al.
Cell, 180(1), 188-204 (2019-12-31)
Glioblastomas exhibit vast inter- and intra-tumoral heterogeneity, complicating the development of effective therapeutic strategies. Current in vitro models are limited in preserving the cellular and mutational diversity of parental tumors and require a prolonged generation time. Here, we report methods for
Fadi Jacob et al.
Cell stem cell, 27(6), 937-950 (2020-10-05)
Neurological complications are common in patients with COVID-19. Although severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), the causal pathogen of COVID-19, has been detected in some patient brains, its ability to infect brain cells and impact their function is not
Tamara Topală et al.
Clujul medical (1957), 87(4), 215-219 (2014-01-01)
The continuous search for new molecules with therapeutic abilities has led to the synthesis and characterization of a large number of metal complexes, proven to exhibit potential as pharmacological agents through their antibacterial, antiviral, antifungal and antineoplastic properties. As serum
Balázs Rada et al.
Journal of immunology (Baltimore, Md. : 1950), 181(7), 4883-4893 (2008-09-20)
The dual oxidase-thiocyanate-lactoperoxidase (Duox/SCN(-)/LPO) system generates the microbicidal oxidant hypothiocyanite in the airway surface liquid by using LPO, thiocyanate, and Duox-derived hydrogen peroxide released from the apical surface of the airway epithelium. This system is effective against several microorganisms that
Eva P Browne et al.
Aquatic toxicology (Amsterdam, Netherlands), 92(4), 281-287 (2009-04-07)
Polybrominated diphenyl ethers (PBDEs) are brominated flame retardants that persist in the environment and are present in geographically widespread fish species. PBDE concentrations can be particularly high in resident Chinook salmon (Onchorhynchus tshawytscha) in the Puget Sound, Washington. Although PBDE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service