Merck
  • ROS-dependent inhibition of the PI3K/Akt/mTOR signaling is required for Oroxylin A to exert anti-inflammatory activity in liver fibrosis.
ROS-dependent inhibition of the PI3K/Akt/mTOR signaling is required for Oroxylin A to exert anti-inflammatory activity in liver fibrosis.
International immunopharmacology (2020-06-09)
Min Shen, Mei Guo, Zhenyi Wang, Yujia Li, Desong Kong, Jiangjuan Shao, Shanzhong Tan, Anping Chen, Feng Zhang, Zili Zhang, Shizhong Zheng
ABSTRACT
More and more evidence showed that autophagy is an inflammation-related defense mechanism against a variety of diseases including liver fibrosis. However, the essential mechanisms remain poorly understood. In this study, we sought to elucidate the impact of Oroxylin A on autophagy and further to identify the potential mechanism of its anti-inflammatory activity. We found that Oroxylin A played a critical role in controlling inflammation in murine liver fibrosis. Moreover, Oroxylin A could inhibit the secretion of pro-inflammatory cytokines in activated hepatic stellate cell (HSCs). We previously reported that Oroxylin A can induce autophagy to alleviate the pathological changes of liver fibrosis and the activation of HSC. Here we further revealed that the inhibition of the PI3K/Akt/mTOR signaling was required for Oroxylin A to induce autophagy activation, which may be the underlying mechanism of the anti-inflammatory activity of Oroxylin A. Interestingly, mTOR overexpression completely impaired the Oroxylin A-mediated autophagy activation, and in turn, damaged the anti-inflammatory activity. Importantly, Oroxylin A inhibited PI3K/Akt/mTOR signaling by scavenging reactive oxygen species (ROS). ROS accumulation by buthionine sulfoximine (BSO) could abrogate the Oroxylin A-mediated ROS elimination, the inhibition of PI3K/Akt/mTOR signaling, and anti-inflammatory activities. Overall, our results provided reliable evidence for the molecular mechanism of Oroxylin A-mediated anti-fibrosis activity, and also identified a new target for drug therapy of liver fibrosis.
Sigma-Aldrich
L-Buthionine-sulfoximine, ≥97% (TLC)
Sigma-Aldrich
Carbon-13C tetrachloride, 99 atom % 13C
Sigma-Aldrich
Human IL18 / Interleukin-18 ELISA Kit, for serum, plasma, cell culture supernatants and urine
Corning® CellBIND® Multiple Well Plate, size 6 wells, flat clear bottom, sterile, lid
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Histodenz, nonionic density gradient medium
Oroxylin A, phyproof® Reference Substance