Skip to Content
Merck
All Photos(1)

Key Documents

488488

Sigma-Aldrich

Carbon-13C tetrachloride

99 atom % 13C

Synonym(s):

Tetrachloromethane-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CCl4
CAS Number:
Molecular Weight:
154.82
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Assay

99% (CP)

refractive index

n20/D 1.46 (lit.)

bp

76-77 °C (lit.)

mp

−23 °C (lit.)

density

1.604 g/mL at 25 °C (lit.)

application(s)

environmental

mass shift

M+1

SMILES string

Cl[13C](Cl)(Cl)Cl

InChI

1S/CCl4/c2-1(3,4)5/i1+1

InChI key

VZGDMQKNWNREIO-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Ozone 1 - STOT RE 1 Inhalation

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Shen et al.
International immunopharmacology, 85, 106637-106637 (2020-06-09)
More and more evidence showed that autophagy is an inflammation-related defense mechanism against a variety of diseases including liver fibrosis. However, the essential mechanisms remain poorly understood. In this study, we sought to elucidate the impact of Oroxylin A on
Zili Zhang et al.
Life sciences, 238, 116934-116934 (2019-10-15)
Proliferation and differentiation of hepatic stellate cells (HSCs) are the most noticeable events in hepatic fibrosis, in which the loss of lipid droplets (LDs) is the most important feature. However, the complex mechanisms of LD disappearance have not been fully
Yujia Li et al.
Free radical biology & medicine, 160, 845-859 (2020-09-19)
Currently, the existing treatments have not cured the liver fibrosis thoroughly. Ferroptosis is a newly discovered way of cell death, which is closely related to many diseases. Previous studies have shown that ferroptosis plays an important role in the occurrence
Zili Zhang et al.
Autophagy, 14(12), 2083-2103 (2018-08-08)
Ferroptosis is a recently recognized form of regulated cell death that is characterized by lipid peroxidation. However, the molecular mechanisms regulating ferroptosis are largely unknown. In this study, we report that the RNA-binding protein ELAVL1/HuR plays a crucial role in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service