Merck
Cue-Evoked Dopamine Promotes Conditioned Responding during Learning.
Neuron (2020-02-07)
Joachim Morrens, Çağatay Aydin, Aliza Janse van Rensburg, José Esquivelzeta Rabell, Sebastian Haesler
ABSTRACT
Dopamine neurons mediate the association of conditioned stimuli (CS) with reward (unconditioned stimuli, US) by signaling the discrepancy between predicted and actual reward during the US. Some theoretical models suggest that learning is also influenced by the salience or associability of the CS. A hallmark of CS associability models is that they can explain latent inhibition, i.e., the observation that novel CS are more effectively learned than familiar CS. Novel CS are known to activate dopamine neurons, but whether those responses affect associative learning has not been investigated. Here, we used fiber photometry to characterize dopamine responses to inconsequential familiar and novel stimuli. Using bidirectional optogenetic modulation during conditioning, we then show that CS-evoked dopamine promotes conditioned responses. This suggests that Pavlovian conditioning is influenced by CS dopamine, in addition to US reward prediction errors. Accordingly, the absence of dopamine responses to familiar CS might explain their slower learning in latent inhibition.
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Sigma-Aldrich
trans-2,cis-6-Nonadienal, mixture of isomers, ≥96%, stabilized, FCC, FG
Sigma-Aldrich
Heptanal, ≥95%, FCC, FG
Sigma-Aldrich
Salicylaldehyde, ≥98%, FG
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
2-Pentenoic acid, predominantly trans, 98%
Sigma-Aldrich
Paraffin oil, puriss., meets analytical specification of Ph. Eur., BP, viscous liquid
Supelco
Thiophene, analytical standard
Sigma-Aldrich
2,3,5-Trimethylpyrazine, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl valerate, ≥98%, FG
Sigma-Aldrich
Thiophene, ≥99%
Sigma-Aldrich
(S)-(−)-Limonene, ≥95%, FG
Sigma-Aldrich
2,3-Dimethylpyrazine, ≥95%, FCC, FG
Sigma-Aldrich
(S)-(+)-Carvone, 96%
Sigma-Aldrich
Isoamyl acetate, ≥95%, FCC, FG
Sigma-Aldrich
Phenethyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
4-Pentenoic acid, ≥98%, stabilized, FG
Sigma-Aldrich
Anisole, ≥99%, FCC, FG
Sigma-Aldrich
α-Phellandrene, natural, ≥85%, FG
Sigma-Aldrich
Benzyl acetate, ≥99%, FCC, FG