Skip to Content
Merck
All Photos(1)

Key Documents

W337706

Sigma-Aldrich

trans-2,cis-6-Nonadienal

mixture of isomers, ≥96%, stabilized, FCC, FG

Synonym(s):

(E,Z)-2,6-Nonadienal, Violet leaf aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH2CH2CH=CHCHO
CAS Number:
Molecular Weight:
138.21
FEMA Number:
3377
EC Number:
Council of Europe no.:
659
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.058
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥96%

contains

α-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.474 (lit.)

bp

94-95 °C/18 mmHg (lit.)

density

0.86 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cucumber; fatty; green; vegetable; violet

SMILES string

[H]C(=O)C(\[H])=C(/[H])CC\C([H])=C(\[H])CC

InChI

1S/C9H14O/c1-2-3-4-5-6-7-8-9-10/h3-4,7-9H,2,5-6H2,1H3/b4-3-,8-7+

InChI key

HZYHMHHBBBSGHB-ODYTWBPASA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-2,cis-6-Nonadienal is the key contributor to the cucumber flavor. It is also found in mango, ripened anchovy and kelp.

Application


  • Induction of a wide range of C(2-12) aldehydes and C(7-12) acyloins in the kidney of Wistar rats after treatment with a renal carcinogen, ferric nitrilotriacetate.: Toyokuni et al. explore the biochemical impact of ferric nitrilotriacetate, a renal carcinogen, on the generation of various aldehydes including trans-2,cis-6-Nonadienal in the kidneys of Wistar rats. This research highlights the role of oxidative stress and lipid peroxidation in renal carcinogenesis (Toyokuni et al., 1997).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

181.4 °F

Flash Point(C)

83 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavor development in the ripening of anchovy (Engraulis encrasicholus L.).
Triqui R & Reineccius GA.
Journal of Agricultural and Food Chemistry, 43(2), 453-458 (1995)
Trans-2, cis-6-nonadienal and trans-2-nonenal in cucumber fruits.
Sekiya J, et al.
Phytochemistry, 16(7), 1043-1044 (1977)
Study of flavor compounds of essential oil extracts from edible Japanese kelps.
Kajiwara T, et al.
Journal of Food Science, 53(3), 960-962 (1988)
Pinar Omür-Ozbek et al.
Environmental science & technology, 45(2), 468-473 (2010-12-15)
Modeling of human exposure to aqueous algal odorants geosmin (earthy), 2-methylisoborneol (musty), and (trans,cis)-2,6-nonadienal (cucumber, fishy), and the solvent trichloroethylene (sweet chemical), was investigated to improve the understanding of water-air transfer by including humans as sensors to detect contaminants. A
Cheryl Palma-Harris et al.
Journal of agricultural and food chemistry, 50(17), 4875-4877 (2002-08-09)
The ability of nonacidified, refrigerated pickled cucumbers to produce the fresh cucumber flavor impact compounds (E,Z)-2,6-nonadienal and (E)-2-nonenal declined during storage. Production of these compounds decreased as the pH of refrigerated cucumbers was reduced. Despite the fact that the concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service