Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium iodide
Linear Formula:
NH
4
I
CAS Number:
12027-06-4
Molecular Weight:
144.94
EC Number:
234-717-7
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (3)
Sustainability
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
ACS reagent (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
221937
Ammonium iodide,
ACS reagent, ≥99%
Expand
View Pricing
203467
Ammonium iodide,
99.999% trace metals basis
Expand
View Pricing
01-4750
Ammonium iodide,
SAJ special grade, ≥99.5%
Expand
View Pricing