About This Item
Recommended Products
grade
ACS reagent
Quality Level
vapor pressure
1 mmHg ( 210.9 °C)
Assay
≥99%
form
powder or crystals
greener alternative product characteristics
Catalysis
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
impurities
≤0.005% insolubles
ign. residue
≤0.05%
pH
4.5-6.5 (25 °C, 50 g/L)
anion traces
Cl- and Br-: ≤0.005%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.05%
cation traces
Ba: ≤0.002%
Fe: ≤5 ppm
heavy metals (as Pb): ≤0.001%
greener alternative category
SMILES string
N.I
InChI
1S/HI.H3N/h1H;1H3
InChI key
UKFWSNCTAHXBQN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
The high yield and regioselective conversion of an unactivated aziridine to an oxazolidinone using carbon dioxide with ammonium iodide as the catalyst
Application
Chemoselective Bromination of Active Methylene and Methyne Compounds by Potassium Bromide, Hydrochloric Acid and Hydrogen Peroxide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT RE 1 Oral
Target Organs
Thyroid
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service