Skip to Content
Merck
All Photos(1)

Key Documents

1706698

USP

Urea

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

urea

manufacturer/tradename

USP

mp

132-135 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

15-25°C

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Urea USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Urea C 13
  • Urea Compounded Irrigation
  • Fluorouracil Cream
  • Allantoin
  • Fluorouracil
Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorouracil
United States Pharmacopeia, 39(3), 1954-1954 (2020)
Allantoin
United States Pharmacopeia, 40(6), 130-130 (2019)
Urea
United States Pharmacopeia, 40(4), 4555-4555 (2020)
Annette Bahlinger et al.
Angewandte Chemie (International ed. in English), 53(33), 8779-8783 (2014-03-20)
β-Amino thioesters are important natural building blocks for the synthesis of numerous bioactive molecules. An organocatalyzed Mannich reaction was developed which provides direct and highly stereoselective access to acyclic β(2)- and β(2,3,3)-amino thioesters with adjacent tertiary and quaternary stereocenters. Mechanistic
A mechanically and electrically self-healing supercapacitor.
Hua Wang et al.
Advanced materials (Deerfield Beach, Fla.), 26(22), 3638-3643 (2014-02-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service