Skip to Content
Merck
All Photos(2)

Documents

15270

Sigma-Aldrich

Biuret

98% (Contains ≤10%(w/w) Triuret)

Synonym(s):

Allophanic acid amide, Carbamoylurea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONHCONH2
CAS Number:
Molecular Weight:
103.08
Beilstein:
1703510
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98% (Contains ≤10%(w/w) Triuret)

form

powder or crystals

impurities

≤1% water

mp

185-190 °C (dec.) (lit.)

SMILES string

NC(=O)NC(N)=O

InChI

1S/C2H5N3O2/c3-1(6)5-2(4)7/h(H5,3,4,5,6,7)

InChI key

OHJMTUPIZMNBFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Biuret (Carbamoylurea) was used as a source of dietary nitrogen in purified diets for steers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Evaluation of urea, biuret, urea phosphate and uric acid as NPN sources for cattle.
R R Oltjen et al.
The Journal of nutrition, 94(2), 193-202 (1968-02-01)
Shuping Jin et al.
Journal of agricultural and food chemistry, 59(1), 322-327 (2010-12-16)
In this investigation, a novel water-insoluble slow-release fertilizer, biuret polyphosphoramide (BPAM), was formulated and synthesized from urea, phosphoric acid (H(3)PO(4)), and ferric oxide (Fe(2)O(3)). The structure of BPAM was characterized by Fourier transform infrared (FTIR) spectroscopy. Subsequently, a coated slow-release
Jerry A Katzmann et al.
Clinical chemistry, 61(2), 360-367 (2014-12-03)
The use of electrophoresis to monitor monoclonal immunoglobulins migrating in the β fraction may be difficult because of their comigration with transferrin and complement proteins. Immunoassays specific for IgGκ, IgGλ, IgAκ, IgAλ, IgMκ, and IgMλ heavy/light chain (HLC) were validated
S J Woltman et al.
Journal of pharmaceutical and biomedical analysis, 14(1-2), 155-164 (1995-12-01)
An electrochemical detection method was applied to the determination of the synthetic peptide TP9201 (Telios Pharmaceuticals). The method utilizes reversed-phase HPLC, followed by post-column formation of Cu(II)-peptide complexes to render peptides electrochemically active via the Cu(III/II) couple. TP9201 is cyclic
Christian Johannessen et al.
Dalton transactions (Cambridge, England : 2003), (10)(10), 1028-1033 (2007-02-28)
Vibrational absorption (VA) and vibrational circular dichroism (VCD) spectroscopy was applied in the analysis of vibrational and low lying electronic transitions of a triplet ground state cobalt(III) coordination compound. The spectroscopic measurements were performed on the tetrabutylammonium salt of (6S,7S)-1,3,5,8,10,12-hexaaza-2,4,9,11-tetraoxo-6,7-diphenyl-dodecanato(4-)cobaltate(III)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service