Skip to Content
Merck
All Photos(1)

Documents

1338007

USP

Imipramine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

10,11-Dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine hydrochloride, 5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenz[b,f]azepine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2 · HCl
CAS Number:
Molecular Weight:
316.87
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

imipramine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].CN(C)CCCN1c2ccccc2CCc3ccccc13

InChI

1S/C19H24N2.ClH/c1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21;/h3-6,8-11H,7,12-15H2,1-2H3;1H

InChI key

XZZXIYZZBJDEEP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Imipramine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Imipramine Hydrochloride Tablets
  • Imipramine Hydrochloride Injection
  • Desipramine Hydrochloride
  • Clomipramine Hydrochloride
  • Desipramine Hydrochloride Tablets
  • Trimipramine Maleate

Biochem/physiol Actions

Tricyclic antidepressant; inhibits the serotonin and norepinephrine transporters with Kis of 7.7 nM and 67 nM, respectively. Has little effect on the dopamine transporter (Ki = 25 μM).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ritabrata Banerjee et al.
Psychiatry investigation, 11(3), 297-306 (2014-08-12)
The present study aimed to investigate whether graded doses of Bacopa Monniera (BM) extract could produce antidepressant-like effects in chronic unpredictable stress (CUS) induced depression in rats and its possible mechanism(s). Rats were subjected to an experimental setting of CUS.
Kazumi Miyawaki et al.
Journal of pharmacological sciences, 127(3), 382-390 (2015-04-04)
Biological rhythms are critical in the etiology of mood disorders; therefore, effective mood disorder treatments should address rhythm disturbances. Among the variables synchronized with the light-dark cycle, spontaneous activity in rodents is useful for investigating circadian rhythms. However, previous studies
Helena Domin et al.
Behavioural brain research, 273, 23-33 (2014-07-22)
The glutamatergic predominance in the excitatory-inhibitory balance is postulated to be involved in the pathogenesis of depression. Such imbalance may be induced by astrocyte ablation which reduces glutamate uptake and increases glutamate level in the synaptic cleft. In the present
Gu Ping et al.
Pharmacology, biochemistry, and behavior, 124, 5-12 (2014-05-23)
Valsartan is a synthetic non-peptide angiotensin II type 1 receptor antagonist that dilates blood vessels and reduces blood pressure by blocking the action of angiotensin, and is safe and well tolerated in hypertensive patients. Population-based studies have suggested a positive
Juliana O G Nascimento et al.
Brain research bulletin, 109, 39-45 (2014-10-08)
The dorsomedial hypothalamus (DMH) has long been associated with the regulation of escape, a panic-related defensive response. Previous evidence has shown that the activation of serotonin (5-HT) 1A and 2A receptors impairs escape behavior induced by the electrical stimulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service