Skip to Content
Merck
All Photos(1)

Documents

I-902

Supelco

Imipramine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2 · HCl
CAS Number:
Molecular Weight:
316.87
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.CN(C)CCCN1c2ccccc2CCc3ccccc13

InChI

1S/C19H24N2.ClH/c1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21;/h3-6,8-11H,7,12-15H2,1-2H3;1H

InChI key

XZZXIYZZBJDEEP-UHFFFAOYSA-N

Gene Information

General description

Imipramine, also known as melipramine, is a tricyclic antidepressant prescribed for the treatment of major depression, enuresis, and anxiety disorder. This certified solution standard is suitable for use in forensic analysis, clinical toxicology, or urine drug testing applications by LC-MS/MS or GC/MS. Imipramine is marketed under the trade name Tofranil.

Application


  • UV-C side-emitting optical fiber-based disinfection: a promising approach for infection control in tight channels.: This study explores the use of UV-C side-emitting optical fibers for disinfection in confined spaces, utilizing PTFE tubing for effective light delivery (Mohsin et al., 2024).

  • Manufacture and Deformation Angle Control of a Two-Direction Soft Actuator Integrated with SMAs.: Development of a two-direction soft actuator using shape memory alloys and PTFE tubing to control deformation angles, highlighting its potential in advanced robotic applications (Acevedo-Velazquez et al., 2024).

  • 3D-printed stereolithographic fluidic devices for automatic nonsupported microelectromembrane extraction and clean-up of wastewater samples.: This study demonstrates the use of PTFE tubing in 3D-printed fluidic devices for efficient microelectromembrane extraction in wastewater treatment (Sahragard et al., 2024).



Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam J Walker et al.
Behavioural brain research, 242, 76-83 (2013-01-02)
Several animal models are currently utilised in the investigation of major depressive disorder; however, each is validated by its response to antidepressant pharmacotherapy. Few animal models consider the notion of antidepressant treatment resistance. Chronic daily administration of adrenocorticotropic hormone (ACTH)
Pablo Méndez et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(47), 16616-16628 (2012-11-24)
Correct brain functioning relies on the precise activity of a myriad of synapses assembling neurons in complex networks. In the hippocampus, highly diverse inhibitory circuits differently govern several physiologically relevant network activities. Particularly, perisomatic inhibition provided by specific interneurons was
Management of nocturnal enuresis.
Patrina H Y Caldwell et al.
BMJ (Clinical research ed.), 347, f6259-f6259 (2013-10-31)
Tatyana Strekalova et al.
Behavioural brain research, 245, 101-106 (2013-02-26)
Tricyclics and selective serotonin reuptake inhibitors (SSRIs) are probably the most widely employed reference antidepressants in animal studies on depression. Using imipramine and citalopram, we sought to assess which drug would be more appropriate as pharmacological reference in paradigms of
Indrek Saar et al.
Journal of neurochemistry, 127(1), 114-123 (2013-04-23)
Neuropeptide galanin and its three G-protein coupled receptors, galanin receptor type 1-galanin receptor type 3 (GalR1-GalR3), are involved in the regulation of numerous physiological and disease processes, and thus represent tremendous potential in neuroscience research and novel drug lead development.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service