Skip to Content
Merck
All Photos(1)

Key Documents

1279000

USP

Fluorouracil

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5-Fluorouracil, 2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1H,3H)-pyrimidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3FN2O2
CAS Number:
Molecular Weight:
130.08
Beilstein:
127172
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

fluorouracil

manufacturer/tradename

USP

mp

282-286 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FC1=CNC(=O)NC1=O

InChI

1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)

InChI key

GHASVSINZRGABV-UHFFFAOYSA-N

Gene Information

human ... TYMS(7298)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluorouracil USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Flucytosine
  • Flucytosine Capsules
  • Fluorouracil
  • Fluorouracil Cream
  • Fluorouracil Injection
  • Fluorouracil Topical Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

S López-Estévez et al.
Gene therapy, 21(7), 673-681 (2014-05-09)
Suicide gene therapy (SGT) is a promising strategy for treating cancer. In this work, we show that thymidine phosphorylase (TP) deficiency, the underlying genetic defect in mitochondrial neurogastrointestinal encephalomyopathy (MNGIE), presents an opportunity to apply SGT using capecitabine, a commonly
Tania Diaz et al.
Journal of surgical oncology, 109(7), 676-683 (2014-02-11)
Surgery is the standard treatment for colorectal cancer (CRC), and adjuvant chemotherapy has been shown to be effective in stage III but less so in stage II. We have analyzed the expression of the miR-200 family in tissue samples from
John M L Ebos et al.
EMBO molecular medicine, 6(12), 1561-1576 (2014-11-02)
Thousands of cancer patients are currently in clinical trials evaluating antiangiogenic therapy in the neoadjuvant setting, which is the treatment of localized primary tumors prior to surgical intervention. The rationale is that shrinking a tumor will improve surgical outcomes and
Steven R Alberts et al.
PharmacoEconomics, 32(12), 1231-1243 (2014-08-27)
Prior economic analysis that compared the 12-gene assay to published patterns of care predicted the assay would improve outcomes while lowering medical costs for stage II, T3, mismatch-repair-proficient (MMR-P) colon cancer patients. This study assessed the validity of those findings
Zhe Guo et al.
Annals of surgical oncology, 21(9), 3069-3076 (2014-04-15)
It is unclear whether hepatic resection (HR) or transarterial chemoembolization (TACE) is associated with better outcomes for patients with hepatocellular carcinoma (HCC) in Barcelona Clinic Liver Cancer (BCLC) stage A. The present study compared survival for patients with BCLC stage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service