Skip to Content
Merck
All Photos(3)

Key Documents

F8423

Sigma-Aldrich

Fluorouracil

meets USP testing specifications

Synonym(s):

5-Fluorouracil, 2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1H,3H)-pyrimidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3FN2O2
CAS Number:
Molecular Weight:
130.08
Beilstein:
127172
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71
Pricing and availability is not currently available.

Agency

USP/NF
meets USP testing specifications

Quality Level

mp

282-286 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

FC1=CNC(=O)NC1=O

InChI

1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)

InChI key

GHASVSINZRGABV-UHFFFAOYSA-N

Gene Information

human ... TYMS(7298)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiyang Chen et al.
Nature communications, 6, 6808-6808 (2015-04-17)
Wild-type p53-induced phosphatase 1 (Wip1) negatively regulates several tumour suppressor and DNA damage response pathways. However, the impact of Wip1 on haematopoietic stem cell (HSC) homeostasis and aging remains unknown. Here we show that Wip1 is highly expressed in HSCs
Julia K J Ahlskog et al.
Bioorganic & medicinal chemistry letters, 19(16), 4851-4856 (2009-07-21)
We describe the synthesis and characterization of two acetazolamide derivatives containing either a charged fluorophore or an albumin-binding moiety, which restrict binding to carbonic anhydrase IX and XII present on tumor cells. In vivo studies showed the preferentially targeting of
Umashankar Das et al.
Journal of medicinal chemistry, 54(9), 3445-3449 (2011-04-01)
Novel 3,5-bis(benzylidene)-1-[3-(2-hydroxyethylthio)propanoyl]piperidin-4-ones (3a-e) display potent cytotoxicity and a preferential lethality toward various neoplasms compared to some normal cells. The corresponding sulfonic acid analogues 5a-e and an isostere 4 demonstrated substantially lower activity. The leads 3d and 3e possess very high
Aurélien Montagu et al.
European journal of medicinal chemistry, 46(2), 778-786 (2011-01-15)
The synthesis and antiviral evaluation of a series of C5-(1,4- and 1,5-disubstituted-1,2,3-triazolo)-nucleoside derivatives is described. The key steps of this synthesis are regioselective Huisgen's 1,3-dipolar cycloaddition, using either copper-catalyzed azide-alkyne cycloaddition (CuAAC) or ruthenium-catalyzed azide-alkyne cycloaddition (RuAAC) under microwave activation.
Umashankar Das et al.
European journal of medicinal chemistry, 42(1), 71-80 (2006-09-26)
The 3,5-bis(arylidene)-4-piperidones 1 contain the 1,5-diaryl-3-oxo-1,4-pentadienyl pharmacophore which is considered to interact at a complementary binding site in susceptible neoplasms. The hypothesis was formulated that the presence of an acyl group attached to the piperidyl nitrogen atom in series 1

Questions

1–3 of 3 Questions  
  1. Why does the 5-Fluorouracil have no effect on my cell line, even at micromolar concentrations that should be more than high enough?

    1 answer
    1. Some cells, and in particular cancer cells, may be resistant to 5-fluorouracil.  Please see this paper Huanwen, W., et al., Molecular Cancer, 8, 125 (2009) which addresses this observation, as well as references 16 and 30 in this publication.If the cells are treated with the closely related gemcitabine (Sigma-Aldrich Product G6423), and cell growth is not inhibited by that drug either, you will have shown that the "problem" is with the cell line, and not with the drug(s).

      Helpful?

  2. Do you have 5-FU (5-Fluorouracil) for injection? If not, how can I prepare it from the powder?

    1 answer
    1. Sigma-Aldrich lists several fluorouracil products, but we do not offer Fluorouracil for injection. Our 5-Fluorouracil (5-FU) products cannot be given to human patients. (Note that although Product F8423 meets USP testing specifications, it is not a pharmaceutical grade or PharmaGrade product.)Having said that, a solution for injection into research animals could be prepared the same way as the Fluorouracil Injection, USP is. According to the drugs.com, "Fluorouracil Injection, USP, an antineoplastic anti-metabolite, is a colorless to yellow aqueous sterile, nonpyrogenic injectable solution for intravenous administration. Each mL contains: 50 mg fluorouracil; pH is adjusted to 8.6 to 9.4 with sodium hydroxide".

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service