Skip to Content
Merck
All Photos(1)

Key Documents

1269265

USP

Famotidine Related Compound F

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-{[2-(Diaminomethyleneamino)thiazol-4-yl}methylthio]propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N4O2S2
CAS Number:
Molecular Weight:
260.34
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

USP

API family

famotidine

form

powder

packaging

pkg of 25 mg

manufacturer/tradename

USP

storage condition

protect from light

color

white

mp

474.8-478.4 °F (246—248°C)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1)CSCCC(=O)O)NC(=N)N

InChI

1S/C8H12N4O2S2/c9-7(10)12-8-11-5(4-16-8)3-15-2-1-6(13)14/h4H,1-3H2,(H,13,14)(H4,9,10,11,12)

InChI key

JEGZXDCDUSGFSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Famotidine Related Compound F (Famotidine Impurity F) is an impurity of famotidine, which is a hydrophilic, cationic, histamine H2 receptor antagonist drug that effectively inhibits gastric acid secretion in humans.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Famotidine Related Compound F can be used as a pharmaceutical primary standard for the determination of the analyte in pharmaceutical formulations by various analytical techniques.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potentiometric determination of famotidine in pharmaceutical formulations
Ayad MM, et al.
Journal of Pharmaceutical and Biomedical Analysis, 29(1-2) (2002)
Famotidine
Pharmacopeia, US
United States Pharmacopeia, 39(1)(1-2), 1815-1815 (2020)
RP-HPLC determination of famotidine and its potential impurities in pharmaceuticals
Helali N, et al.
Chromatographia, 60(7-8) (2004)
Photoaffinity labeling of the anionic sites in Caco-2 cells mediating saturable transport of hydrophilic cations ranitidine and famotidine
Bourdet DL, et al.
Journal of medicinal chemistry, 47(11) (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service