Skip to Content
Merck
All Photos(1)

Documents

1269221

USP

Famotidine Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N,N′′′-[(1,1-Dioxido-2H-1,2,4,6-thiatriazine-3,5-diyl)bis(2,1-ethanediylthiomethylene-4,2-thiazolediyl)]bisguanidine, 3,5-Bis[2-[[[2-[(diaminomethylene)amino]thiazol-4-yl]methyl]sulphanyl]ethyl]-4H-1,2,4,6-thiatriazine 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23N11O2S5
CAS Number:
Molecular Weight:
561.75
UNSPSC Code:
41116107
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

USP

API family

famotidine

form

powder

packaging

pkg of 25 mg

manufacturer/tradename

USP

storage condition

protect from light

color

yellow to white

solubility

water: soluble

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C16H23N11O2S5/c17-13(18)24-15-21-9(7-32-15)5-30-3-1-11-23-12(27-34(28,29)26-11)2-4-31-6-10-8-33-16(22-10)25-14(19)20/h7-8H,1-6H2,(H,23,26,27)(H4,17,18,21,24)(H4,19,20,22,25)

InChI key

NRRXZFHNRLQONH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Famotidine Related Compound B (Famotidine Impurity B) is an impurity of famotidine, which is a hydrophilic, cationic, histamine H2 receptor antagonist drug that effectively inhibits gastric acid secretion in humans.

Application

Famotidine Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Famotidine

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service