Skip to Content
Merck
All Photos(1)

Key Documents

71023AST

Supelco

Astec® CHIRALDEX B-PH Capillary GC Column

L × I.D. 30 m × 0.25 mm, df 0.12 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
NACRES:
SB.54

material

fused silica

Quality Level

description

GC capillary column

packaging

pkg of 1 ea

parameter

-10-200 °C temperature (isothermal)
-10-220 °C temperature (programmed)

Beta value

500

df

0.12 μm

technique(s)

gas chromatography (GC): suitable

L × I.D.

30 m × 0.25 mm

matrix active group

non-bonded; (S)-2-hydroxy propyl methyl ether derivative of β-cyclodextrin phase

application(s)

agriculture
chemicals and industrial polymers
cleaning products
clinical
cosmetics
environmental
flavors and fragrances
food and beverages
forensics and toxicology
life science and biopharma
personal care
pharmaceutical (small molecule)

column type

capillary chiral

Looking for similar products? Visit Product Comparison Guide

General description

CHIRALDEX B-PH shows at least some selectivity to a great variety of analytes, but is especially effective for saturated analytes with minimal functionality, saturated cyclics and bicyclics. The CHIRALDEX PH series of columns shows less of a necessity for inclusion complexation for chiral recognition than the DA columns. This phase often shows a reversal of elution order (enantioreversal) compared to the B-DA phase.

Chem/Phys Resistance

Temp. Limits:
  • -10 °C to 200 °C isothermal, 220 °C programmed

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective conjugate addition with chiral amidocuprates, part III. Studies on the synthetic optimization of MAPP-cuprates.
Rossiter, Bryant E., et al.
Tetrahedron Asymmetry, 3 (2), 231-234 (1992)
Asymmetric reductions in aqueous media: enzymatic synthesis in cyclodextrin containing buffers
Zelinski, Thomas, et al.
Tetrahedron Asymmetry, 10 (9), 1681-1687 (1999)
Alkali metal fluorides as efficient fluorinating agents. Enantiocontroued synthesis of 2-fluoroalkyl carboxylates and 1-fluoroalkyl benzenes
Fritz-Langhals, Eike
Tetrahedron Asymmetry, 5 (6), 981-986 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service