Skip to Content
Merck
All Photos(2)

Documents

270849

Sigma-Aldrich

2-Naphthalenethiol

≥95%

Synonym(s):

2-Naphthyl mercaptan, Thio-2-naphthol (β)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7SH
CAS Number:
Molecular Weight:
160.24
Beilstein:
636389
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

bp

286 °C (lit.)

mp

79-81 °C (lit.)

solubility

diethyl ether: very soluble(lit.)
ethanol: very soluble(lit.)
petroleum ether: very soluble(lit.)
water: slightly soluble(lit.)

SMILES string

Sc1ccc2ccccc2c1

InChI

1S/C10H8S/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7,11H

InChI key

RFCQDOVPMUSZMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The magnetic resonance shift for a self-assembled monolayer of 2-naphthalenethiol was studied that suggested considerable promise in flexible and transparent photonic devices for biological and chemical sensing.

Application

2-Naphthalenethiol was used in the preparation of cholesterol monolayer and multilayer Langmuir- Blodgett (LB) films. The electrochemical barrier properties of these films were studied using cyclic voltammetry and electrochemical impedance spectroscopy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinlong Xu et al.
Nano letters, 11(8), 3232-3238 (2011-06-24)
Flexible electronic and photonic devices have been demonstrated in the past decade, with significant promise in low-cost, light-weighted, transparent, biocompatible, and portable devices for a wide range of applications. Herein, we demonstrate a flexible metamaterial (Metaflex)-based photonic device operating in
R A Alvarez-Puebla et al.
The Analyst, 129(12), 1251-1256 (2004-11-27)
The results of the search for the optimal experimental conditions for ultrasentitive chemical analysis of 1-naphthalenethiol (1-NAT) and 2-naphthalenethiol (2-NAT) using surface-enhanced Raman scattering (SERS) are discussed. The report begins with a review of the vibrational spectra, including infrared and
Metal-cluster catalysts: Access granted.
Graham J Hutchings
Nature chemistry, 2(12), 1005-1006 (2010-11-26)
Namal de Silva et al.
Nature chemistry, 2(12), 1062-1068 (2010-11-26)
In enzymes, the electronic and steric environments of active centres, and therefore their activity in biological processes, are controlled by the surrounding amino acids. In a similar manner, organic ligands have been used for the 'passivation' of metal clusters, that
Bong-Hyun Jun et al.
Analytical biochemistry, 391(1), 24-30 (2009-05-13)
This article presents a prototype of a surface-enhanced Raman spectroscopy (SERS)-encoded magnetic bead of 8mum diameter. The core part of the bead is composed of a magnetic nanoparticle (NP)-embedded sulfonated polystyrene bead. The outer part of the bead is embedded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service