Skip to Content
Merck
All Photos(1)

Key Documents

64843

Supelco

Celite® R566 Inorganic Adsorbent

filter aid

Synonym(s):

Celite 566

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
SB.52

Product Name

Celite® R566,

grade

reagent grade

form

powder

technique(s)

LPLC: suitable

surface area

500 m2/g , approx.

matrix

Silica

matrix active group

silica

particle size

2000 μm

pore size

120 Å mean pore size

separation technique

hydrophilic interaction (HILIC)

General description

Celite® R566 is diatomaceous earth used to filter analytes from the matrices.
Celite® R566 is primarily used to interact with target reagents, but it may also be adjusted to fit corresponding scientific methods.

Application

Celite® R566 can be used:
  • As an adsorbent in the Soxhlet method to determine the fat content of lupin-based milk alternatives (LMAs)
  • For filtration of the crude reaction mixture in the synthesis of novel fluorinated and non-fluorinated di(thiazolyl)benzenes
  • To filter the polymeric reaction mixtures in the study to evaluate the effect of lipids on cold crystallization of natural rubber
  • As a filter in the synthesis of 1,4-cis-poly (isoprene) macroinitiators from natural rubber

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New insight into the cold crystallization of natural rubber: The role of linked and free fatty chains
Grange J, et al.
Polymer Crystallization, 2(4), e10075-e10075 (2019)
Physicochemical and nutritional properties of high protein emulsion-type lupin-based model milk alternatives: effect of protein source and homogenization pressure
Vogelsang-O'Dwyer M, et al.
Journal of the Science of Food and Agriculture (2021)
Facile synthesis of 1, 4-cis-polyisoprene-polypeptide hybrids with different architectures
Radchenko A, et al.
Polym. Chem., 10(19), 2456-2468 (2019)
Synthesis, characterization and crystal structures of novel fluorinated di (thiazolyl) benzene derivatives
Barlog M, et al.
Organic Chemistry Frontiers : An International Journal of Organic Chemistry / Royal Society of Chemistry, 6(6), 780-790 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service