717185
Silica gel Inorganic Sorbent
technical grade, 230-400 mesh
About This Item
Recommended Products
Product Name
Silica gel, technical grade, pore size 60 Å, 230-400 mesh particle size, 40-63 μm particle size
grade
technical grade
technical (grade)
Quality Level
form
powder
manufacturer/tradename
Sigma-Aldrich
technique(s)
LPLC: suitable
surface area
≥480 m2/g , approx.
matrix
Silica
matrix active group
silica
particle size
230-400 mesh
40-63 μm
pore size
0.7-0.85 cm3/g pore volume (approx.)
60 Å
bp
2230 °C
mp
>1600 °C
separation technique
hydrophilic interaction (HILIC)
SMILES string
O=[Si]=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service