575653-U
SPE 96-Deep Square Well Collection Plate
well volume 2 mL, polypropylene, pk of 50
Synonym(s):
SPE 96-Well Collection Plate, 2mL, pk 50
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115854
NACRES:
NB.21
Recommended Products
material
polypropylene
packaging
pk of 50
technique(s)
solid phase extraction (SPE): suitable
well volume
2 mL
application(s)
food and beverages
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service