55263-U
Protein Precipitation Filter 96-well solid phase extraction (SPE) Plate
Volume 2 mL, pk of 1
Synonym(s):
Protein Precipitation Plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115712
NACRES:
NB.21
Recommended Products
Product Name
96-Well Protein Precipitation Filter Plate, pk of 1
packaging
pk of 1
technique(s)
solid phase extraction (SPE): suitable
volume
2 mL
General description
The 96-well protein precipitation filter plate is ideal for removing precipitated proteins from biological plasma/serum. The plate consists of a 0.2 μm hydrophobic graded filter/frit. Biological plasma is first added to the 96-well plate followed by a protein precipitating agent (e.g., acetonitrile). After a brief mixing step, vacuum is applied to the plate, and the filter/frit removes precipitated proteins from the sample. The resulting filtrate is ready for further processing and/or analysis.
Application
- Determination of estrone sulfate, testosterone, androstenedione, DHEAS, cortisol, cortisone, and 17α-hydroxyprogesterone by LC-MS/MS in children and adolescents.: This study employs 96-Well Protein Precipitation Filter Plates for rapid sample preparation, significantly increasing the throughput of hormone analysis in pediatric populations. This technique enhances the accuracy and efficiency of hormonal assessments, crucial for clinical and research settings (Ankarberg-Lindgren et al., 2020).
- Quantitative Determination of Ethyl Glucuronide and Ethyl Sulfate in Postmortem and Antemortem Whole Blood Using Phospholipid Removal 96-Well Plate and UHPLC-MS-MS.: This research demonstrates the utility of 96-Well Filter Plates in removing phospholipids from blood samples, streamlining the detection of ethyl glucuronide and ethyl sulfate, substances important for forensic and clinical toxicology (Sidqey et al., 2021).
- Development of a CDC-certified total testosterone assay for adult and pediatric samples using LC-MS/MS.: The study highlights the application of 96-Well Protein Precipitation Filter Plates in developing a CDC-certified assay for testosterone, showing their critical role in enhancing diagnostic accuracy in clinical settings (Star-Weinstock et al., 2019).
- Validation and application of a liquid chromatography-tandem mass spectrometric method for quantification of the drug transport probe fexofenadine in human plasma using 96-well filter plates.: This paper validates a high-throughput method for the analysis of fexofenadine, showcasing the 96-Well Filter Plates′ efficiency in clinical pharmacology and bioanalysis settings (Stanton et al., 2010).
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Miau-Guo Kang et al.
Journal of analytical toxicology, 43(2), 96-103 (2018-10-03)
Benzodiazepines is a large drug group used extensively to treat sleep disorders and anxiety; these drugs are frequently associated with various crimes such as murder and drug-facilitated sexual assault. With growing use and misuse of these compounds, confirmatory assays are
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service