57265
Visiprep™ SPE Vacuum Manifold
DL (Disposable Liner), 24-port model
All Photos(1)
About This Item
UNSPSC Code:
12000000
Recommended Products
material
PTFE frit
polypropylene tube
Quality Level
product line
Visiprep™
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
General description
SPE is a form of chromatography, and, as with other chromatographic techniques, control of flow rate is critical for maintaining reproducible extractions. Unlike other vacuum manifolds, the Visiprep system contains a patented valve system that allows for precise flow control through each SPE tube via rotating, independent, screw-type valves situated in each port within the manifold cover. Visiprep vacuum manifolds allow you to process up to 12 (12-port version) or 24 (24-port version) SPE samples simultaneously. Spacing between ports are slightly smaller on the 24-port versions than the 12-port versions so consideration of tube sizes is recommended.
The Visiprep DL (Disposable Liner) Vacuum Manifold eliminates the possibility of cross-contamination when processing a new sample on the same port. The liner consists of a PP female luer hub that attaches to the SPE, and thin-walled PTFE tubing that is threaded through the SPE port. This ensures that all SPE port and valve surfaces coming in contact with the sample can be replaced following each extraction.
Application
Visiprep™ SPE Vacuum Manifold was used to perform solid phase extraction to separate polyphenols from grapes, in order to determine the polyphenol content using LC-MS analysis. It was also used for concentration and purification of solvent extracts from surface water and sewage water sediments. Furthermore it was also used to extract and determine estrone in water using solid-phase extraction-enzyme-linked immunosorbent assay (SPE-ELISA).
Features and Benefits
- Patented screw-type valves within each SPE port for precise flow control
- Glass basin will not dissolve, fog, or discolor when exposed to solvents
- Legs on cover allow user to easily rest cover on work surface when removed from the manifold
- Screw-type solvent-resistant vacuum bleed gauge and valve offer better sealing and vacuum control. Valve takes 1/4″ vacuum tubing.
- PP collection vessel rack accommodates autosampler vials; small scintillation vials (22.75 mm O.D. recommended); 10 and 16 mm test tubes; and 1, 2, 5, and 10 mL volumetric flasks. An optional plate for 20 mL scintillation vials is available for 12-port models.
Supplied with 100 disposable liners (Part No. 57059).
Legal Information
Visiprep is a trademark of Sigma-Aldrich Co. LLC
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
Guangdi Wang et al.
Journal of environmental monitoring : JEM, 14(5), 1353-1364 (2012-03-23)
Endocrine disrupting compounds (EDCs), represented by steroid hormones, organochlorine pesticides (OCPs), polychlorinated biphenyls (PCBs), and bisphenol A have been determined in four sediment cores from the Gulf of Mexico, from New Orleans surface water (Lake Pontchartrain and Mississippi River), and
Angeles M Alonso Borbalán et al.
Journal of chromatography. A, 1012(1), 31-38 (2003-09-26)
There is considerable interest in the analysis of phenolic compounds due to their involvement in the organoleptic characteristics of foods and drinks, and their contribution to protection against cardiovascular diseases and cancers through their antioxidant activity. This article describes studies
Development of a solid-phase extraction-enzyme-linked immunosorbent assay method for the determination of estrone in water.
Li, Zhilan, et al.
Analytica Chimica Acta, 503, 171-177 (2004)