Skip to Content
Merck
All Photos(1)

Key Documents

57047

Supelco

Visiprep SPE Manifold Replacement Solvent Guide Needles

for use with standard, 12- and 24-port models (included with 57030-U & 57250-U)), PTFE, pk of 12

Synonym(s):

PTFE SOL.GUIDE NEEDLES PK/12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
Pricing and availability is not currently available.

material

PTFE

Quality Level

product line

Visiprep

packaging

pk of 12

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

compatibility

for use with standard, 12- and 24-port models (included with 57030-U & 57250-U))

Looking for similar products? Visit Product Comparison Guide

General description

The manifold lids are equipped with solvent guide needles which provide an inert flow path for samples.

Footnote

Included with Visiprep 12- and 24-port Vacuum Manifolds (57030-U and 57250-U)

Legal Information

Visiprep is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service