Skip to Content
Merck
All Photos(1)

Documents

57089

Supelco

Supelclean LC-4 SPE Tube

Wide Pore, bed wt. 500 mg, volume 3 mL, pk of 54

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712

material

PE frit (20 μm porosity)
polypropylene hardware

Quality Level

product line

Supelclean

feature

endcapped

composition

bed wt., 500 mg

packaging

pk of 54

technique(s)

solid phase extraction (SPE): suitable

volume

3 mL

matrix

silica gel base material (irregularly shaped)

matrix active group

C4 (butyl) bonding

particle size

45 μm

pore size

500 Å pore size

application(s)

food and beverages

separation technique

reversed phase

General description

  • Larger pore size to accommodate larger macromolecules (e.g. proteins and peptide)
  • Commonly used for desalting and extracting proteins/peptides in aqueous samples

Legal Information

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service