Skip to Content
Merck
All Photos(1)

Key Documents

505285

Supelco

LpDNPH Ozone Scrubber

S10L (Reversible Cartridge), Potassium Iodide (KI) 1.5 g, pkg of 10 ea

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12190000

Agency

ASTM® D5197
EPA 100,IP-6A,TO-11A,TO-11A
NIOSH 2016

form

solid

composition

Potassium Iodide (KI), 1.5 g

packaging

pkg of 10 ea

technique(s)

active air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

Ozone scrubbers are recommended for sampling carbonyls in high humidity environments. These cartridges contain 1.5 g of high purity potassium iodide (KI) and are available in two cartridge styles. KI traps the ozone which causes a negative formaldehyde interference in DNPH coated devices. The reversible cartridge offers a slip luer end-fitting, and the rezorian cartridge offers a luer lock end-fitting, to enable you to connect your cartridge of choice directly to the inlet of any DNPH cartridge with a like end-fitting. Testing (200 ppb ozone, 50 % RH, 25°C) indicates the ozone scrubber to have an ozone capacity of 100,000 ppb/hr.

Application

LpDNPH Ozone Scrubber was used to remove ozone, in an experiment conducted to develop emission factors for air pollutants, after burning sugarcane, using combustion chamber method. It was also used to chemically remove ozone and reduce the decomposition, in an experiment conducted to study different active sampling procedures and analysis of aldehydes done at environment level.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PAHs, carbonyls, VOCs and PM 2.5 emission factors for pre-harvest burning of Florida sugarcane.
Hall, Danielle, et al.
Atmospheric Environment, 55, 164-172 (2012)
Cong-Khanh Huynh et al.
Analytical and bioanalytical chemistry, 372(5-6), 654-657 (2002-04-10)
Within the framework of a European interlaboratory exercise, the Vito facility for the generation of controlled atmospheres was used to test the suitability of four sampling techniques for priority aldehydes namely formaldehyde, acrolein, acetaldehyde, and glutaraldehyde at the ranges of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service