Skip to Content
Merck
All Photos(1)

Key Documents

22316-U

Hand crimper, adjustable

Hand crimper, adjustable, for use with 20 mm crimp seals, pkg of 1 ea

Synonym(s):

Adjustable Vial Crimper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

packaging

pkg of 1 ea

compatibility

for use with 20 mm crimp seals

Looking for similar products? Visit Product Comparison Guide

General description

The adjustable hand crimper is ideal if you use varying thickness of septa in your analysis. This product has the ability to be adjusted for changes in septa thickness.
This product is very useful in SPME applications recommending the use of a thinner septa (1.0-1.5mm thickness).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service