Z114146
Wheaton closed-top seals
Tear-off seals only, diam. 20 mm, (no septa)
Synonym(s):
Wheaton Vial Seals
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
Recommended Products
material
(no septa)
feature
removable center
packaging
pkg of 100 ea
diam.
20 mm
compatibility
for use with 20 mm crimp seal vial
Looking for similar products? Visit Product Comparison Guide
General description
It is used for sealing vials, bottles, etc. It is air-tight, and prevents contamination.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service