Skip to Content
Merck
All Photos(1)

Key Documents

20744AST

Supelco

Astec® CYCLOBOND I 2000 DMP Chiral (5 μm) HPLC Columns

L × I.D. 25 cm × 21.2 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

Astec® CYCLOBOND I 2000 DMP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm

material

stainless steel column

Quality Level

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-50 °C temperature
172 bar pressure (2500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

25 cm × 21.2 mm

matrix

silica particle platform
fully porous particle

matrix active group

cyclodextrin, beta- phase

particle size

5 μm

pore size

100 Å

operating pH

3.5-7

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

The reaction of the 3,5-dimethylphenyl isocyanate with the hydroxyl groups of β-cyclodextrin results in a pi-basic phase similar in character to the naphthylethyl carbamate phases. The selectivity is greater for the CYCLOBOND I 2000 DMP when the chiral center of the analyte is part of a ring structure or is on the α carbon. This phase has been very useful for derivatized amines, like amphetamine ACQ.

  • Bonded phase: 3,5-Dimethylphenyl carbamate modified β-cyclodextrin

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reprint of: Enantiomeric separation of functionalized ethano-bridged Troger bases using macrocyclic cyclofructan and cyclodextrin chiral selectors in high-performance liquid chromatography and capillary electrophoresis with application of principal component analysis
Weatherly, Choyce A., et al.
Journal of Chromatography. B, Biomedical Applications, 968, 40-48 (2014)
Enantiomeric separation of functionalized ethano-bridged Troger bases using macrocyclic cyclofructan and cyclodextrin chiral selectors in high-performance liquid chromatography and capillary electrophoresis with application of principal component analysis
Weatherly, Choyce A., et al.
Journal of Chromatography. B, Biomedical Applications, 955-956, 72-80 (2014)
István Ilisz et al.
Chirality, 21(3), 339-348 (2008-06-17)
The application of 3,5-dimethylphenyl-carbamoylated-beta-cyclodextrin (Cyclobond I 2000 DMP) and 2,6-dinitro-4-trifluoromethylphenyl-ether-beta-cyclodextrin-based (Cyclobond DNP) chiral stationary phases for the high-performance liquid chromatographic enantioseparation of unusual beta-amino acids is reported. The investigated amino acids were saturated or unsaturated alicyclic beta-3-homo-amino acids and bicyclic
Nilusha L T Padivitage et al.
Drug testing and analysis, 6(6), 542-551 (2013-10-12)
Recently a novel class of chiral stationary phases (CSPs) based on cyclofructan (CF) has been developed. Cyclofructans are cyclic oligosaccharides that possess a crown ether core and pendent fructofuranose moieties. Herein, we evaluate the applicability of these novel CSPs for
Xinxin Han et al.
Journal of chromatography. A, 1063(1-2), 111-120 (2005-02-11)
The enantiomeric separation of a set of 30 new chiral furan derivatives has been achieved on native and derivatized beta-cyclodextrin stationary phases using high performance liquid chromatography (HPLC). The hydroxypropyl-beta-cyclodextrin (Cyclobond RSP), the 2,3-dimethyl-beta-cyclodextrin (Cyclobond DM), and the acetyl-beta-cyclodextrin (Cyclobond

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service