Merck
  • Separation of chiral furan derivatives by liquid chromatography using cyclodextrin-based chiral stationary phases.
Separation of chiral furan derivatives by liquid chromatography using cyclodextrin-based chiral stationary phases.
Journal of chromatography. A (2005-02-11)
Xinxin Han, Tuanli Yao, Ying Liu, Richard C Larock, Daniel W Armstrong
ABSTRACT
The enantiomeric separation of a set of 30 new chiral furan derivatives has been achieved on native and derivatized beta-cyclodextrin stationary phases using high performance liquid chromatography (HPLC). The hydroxypropyl-beta-cyclodextrin (Cyclobond RSP), the 2,3-dimethyl-beta-cyclodextrin (Cyclobond DM), and the acetyl-beta-cyclodextrin (Cyclobond AC) stationary phases are the most effective chiral stationary phases (CSPs) for the separation of these racemates in the reverse phase mode. No enantioseparations have been observed on the native beta-cyclodextrin chiral stationary phase (Cyclobond I 2000) and only a few separations have been attained on the S-naphthylethyl carbamate beta-cyclodextrin (Cyclobond SN) and 3,5-dimethylphenyl carbamate beta-cyclodextrin (Cyclobond DMP) chiral stationary phases in the reverse phase mode. The polar organic and the normal phase mode on these CSPs are not effective for separation of these compounds. The characteristics of the analytes, including steric bulk, hydrogen bonding ability, and geometry, play an important role in the chiral recognition process. The pH affects the enantioseparation of compounds with ionizable groups and the addition of 0.5% methyl tert-butyl ether to the mobile phase significantly enhances the separation efficiency for some highly retained compounds.
Sigma-Aldrich
Acetyl-β-cyclodextrin, Monoacetyl-β-cyclodextrin
Supelco
Astec® CYCLOBOND I 2000 HP-RSP Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Astec® CYCLOBOND I 2000 HP-RSP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CYCLOBOND I 2000 DMP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CYCLOBOND I 2000 AC Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CYCLOBOND I 2000 DMP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Astec® CYCLOBOND I 2000 DMP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm