20563
Glass Cartridge and Screen for Large PUF Plug
pkg of 1 ea
Synonym(s):
Large PUF Cartridges
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12190000
Recommended Products
material
glass cartridge
packaging
pkg of 1 ea
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
compatibility
for use with ORBO-2000 (6.5 cm Large PUF Plug)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service