Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Recommended Products
Agency
suitable for ASTM® D4861
suitable for EPA Method 10A, Method 23 and OTM-45
form
solid
packaging
pkg of 100 g
technique(s)
HPLC: suitable
LPLC: suitable
surface area
~300 m2/g
matrix
styrene-divinylbenzene (macroreticular)
matrix active group
polymer
particle size
20-60 mesh
pore size
~0.65 mL/g pore volume
90 Å mean pore size
density
1.02 g/mL at 25 °C (true wet)(lit.)
1.08 g/mL at 25 °C (skeletal)(lit.)
separation technique
reversed phase
storage temp.
2-8°C
SMILES string
N([C@@H](CCC(=O)OC(C)(C)C)C=O)C(=O)OCC1c2c(cccc2)c3c1cccc3
InChI
1S/C24H27NO5/c1-24(2,3)30-22(27)13-12-16(14-26)25-23(28)29-15-21-19-10-6-4-8-17(19)18-9-5-7-11-20(18)21/h4-11,14,16,21H,12-13,15H2,1-3H3,(H,25,28)/t16-/m0/s1
InChI key
BCIPGSZQUDLGSY-INIZCTEOSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Specifications:
Application
- Amberlite™ XAD®-2 was used as adsorbent in the extraction of organochlorine compounds using Soxhlat extraction technique.[3]
- Amberlite™ XAD®-2 was used as adsorbent in the extraction of honey samples using column chromatography for HPLC analysis.[4]
- Amberlite™ XAD®-2 was used in adsorption chromatography.[5]
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the Department of Transportation shipping information for this product?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service