Recommended Products
form
solid
autoignition temp.
800 °F
packaging
pkg of 500 g
technique(s)
HPLC: suitable
LPLC: suitable
surface area
750 m2/g
matrix
styrene-divinylbenzene
matrix active group
polymer
particle size
20-60 mesh
pore size
~0.98 mL/g pore volume
100 Å mean pore size
density
1.02 g/mL (true wet)(lit.)
1.08 g/mL (skeletal)(lit.)
separation technique
reversed phase
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Amberlite XAD4, a nonpolar absorbent, used for stripping and concentration of organic compounds. It is also used in developing chelating ligands due to its properties like, porosity, high surface area, purity and durability.
Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
Application
Amberlite™ XAD4 was used:
- in rapid fractionation technique
- as sorbent for HPLC analysis
Polyaromatic adsorbent for small hydrophobic compounds, surfactants, pharmaceutical manufacturing, phenols, chlorinated organics, pesticide removal and recovery, and organic removal from aqueous food streams.
Other Notes
Two particle sizes are available:
Mesh 20-60 are Cat. Nos. 20276, 10358, XAD4
Mesh 20-50 is Cat. No. 06444
Mesh 20-60 are Cat. Nos. 20276, 10358, XAD4
Mesh 20-50 is Cat. No. 06444
Legal Information
Amberlite is a trademark of DuPont de Nemours, Inc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Characterization and applications of Amberlite XAD-4 in preparative liquid chromatography
Pietrzyk, D. J., and J. D. Stodola
Analytical Chemistry, 53, 1822-1828 (1981)
Mehmet Dogru et al.
Journal of hazardous materials, 149(1), 166-173 (2007-05-01)
The present work proposes the use of Bacillus subtilis immobilized on Amberlite XAD-4 as new biosorbent in trace metal determination. The procedure is based on the biosorption of Cu and Cd ions on a column of Amberlite XAD-4 resin loaded
Imam Bakhsh Solangi et al.
Journal of hazardous materials, 171(1-3), 815-819 (2009-07-18)
Fluoride in drinking water above permissible level is responsible for human being affected by skeletal fluorosis. In this study, Amberlite XAD-4 has been modified by introducing amino group onto the aromatic ring for its application in fluoride remediation. Characterization of
Corjan van den Berg et al.
Biotechnology and bioengineering, 100(3), 466-472 (2008-04-29)
The sustainable production of fine/bulk chemicals is often hampered by product toxicity and inhibition to the producing micro-organisms. Consequently, the product must be removed from the micro-organisms' environment. To achieve this, so-called solvent-impregnated resins (SIRs) as well as commercial resins
Chao Huang et al.
New biotechnology, 29(3), 372-378 (2011-04-05)
Oleaginous yeast Trichosporon fermentans was proved to be able to use sulphuric acid-treated sugar cane bagasse hydrolysate as substrate to grow and accumulate lipid. Activated charcoal was shown as effective as the more expensive resin Amberlite XAD-4 for removing the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service