20267-U
ORBO™ 32 Small Activated Coconut Charcoal (20/40), 100/50 mg
W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Synonym(s):
Activated Coconut Charcoal Tube
About This Item
Recommended Products
material
W,F,F separators
Quality Level
Agency
ASTM® D3686
NIOSH 1000,1001,1002,1003,1004,1005,1006,1008,1010,1011,1012,1014,1015,1016,1017,1018,1020,1022,1300
NIOSH 1301,1302,1400,1401,1402,1403,1404,1450,1451,1452,1454,1457,1458,1459,1500,1501,1550,1551,1552
NIOSH 1600,1602,1603,1604,1608,1609,1610,1611,1612,1613,1616,1617,1618,1619,1620,6001
OSHA 2,3,5,7,8,9,11,12,14,37,48,66,79,83,99,101,111,1002
product line
ORBO™
composition
Bed A, 100 mg
Bed B, 50 mg
packaging
pkg of 50 ea
manufacturer/tradename
ORBO™ 32 Small
technique(s)
active air sampling: suitable
O.D. × L
6 mm × 75 mm
matrix
activated coconut charcoal
activated coconut charcoal
particle size
20-40 mesh
application(s)
air monitoring
environmental
industrial hygiene
compatibility
mode of use Solvent desorption
technique used for Air monitoring
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service