Z732117
Brightmax™ Sealing Films
for luminescence and microscopy, non-sterile
Synonym(s):
immunofluorescence sealing film, microplate sealing film, multiwellplate sealing film
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
Recommended Products
material
vinyl film (vinyl/adhesive)
sterility
non-sterile
packaging
pack of 50 ea
manufacturer/tradename
Excel Scientific WT-50
technique(s)
immunofluorescence: suitable
L
116 mm
W × L
7.6 cm × 11.6 cm
W
76 mm
color
white
General description
BrightMax™ sealing films are 200 μm thick white vinyl adhesive sealing films for use with white microplates. The sealing films may be applied to either the top or bottom of the plate. A split backing aids in applying the sealing film properly. The light-reflective properties of BrightMax™ plate-sealing films on clear-bottom plates maximize sensitivity of bioluminescence and chemiluminscence assays in bottom-reading luminometers. White films also have application as a backing matrix for microscopy of punchout samples from filter plates, for example in ELISPOT assays. Dimensions are 76 by 116 mm with no end tabs. A backing split singly on the long dimension aids in applying film to plates. Recommended temperature range: -40 °C to +80 °C.
- White light-reflecting sealing films for use with white microplates
- May be applied to top or bottom of plate
- Maximize intensity and signal-to-noise ratios in luminescence assays
- Additional applications for mounting and storage of filters for microscopy, for example in ELISPOT assays
Legal Information
Brightmax is a trademark of Excel Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service