Skip to Content
Merck
All Photos(1)

Key Documents

Y2001

Sigma-Aldrich

Yeast Synthetic Drop-out Medium Supplements

without histidine, leucine, tryptophan and uracil

Synonym(s):

Synthetic drop-out medium for yeast

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12000000
NACRES:
ND.02
Pricing and availability is not currently available.

Quality Level

form

powder

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

Application

The selection of plasmids in yeast is based on the use of auxotrophic mutant strains that cannot grow without a specific media component (an amino acid, purine or pyrimidine). Transformation with a plasmid containing the mutated gene enables the transformant to grow on a medium lacking the required component. Sigma′s Yeast Synthetic Drop-Out Media Supplements create a richer medium for better yield and growth rate, and increase the probability of successful transformations when screening libraries or performing gene knock-outs.

Components

The supplements contain all of the following components except as indicated:
Amino acids: All standard amino acids except for histidine, leucine, tryptophan and uracil. Each amino acid is present at a concentration of 76 mg/L.
Other nutrients: Adenine (18 mg/L), inositol (76 mg/L), p-aminobenzoic acid (8 mg/L)

Other Notes

Mixtures of amino acids and other nutrients to be added to Yeast Nitrogen Base Without Amino Acids.

Quantity

1.4 g will supplement 1 liter of medium.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margot Bärenstrauch et al.
Fungal genetics and biology : FG & B, 139, 103383-103383 (2020-04-10)
Plants comprise many asymptomatic fungal endophytes with potential roles of plant protection against abiotic and biotic stresses. Endophytes communicate with their host plant, with other endophytes and with invading pathogens but their language remains largely unknown. This work aims at

Questions

  1. It says to add 1.4g to a liter of medium. Would this mean you would need to add 14g to a 10X stock solution that is suggested when preparing yeast nitrogen base without AA.

    1 answer
    1. Mathematically, yes, this would mean 14 grams would be added to a 10X stock. However, studies of the solubility of this supplement at 10X have not been performed. The end user should attempt this using a smaller volume to verify solubility.
      Please see the links below to the product datasheet, the Yeast Drop Out page, and the Yeast Growth Protocols page:

      Y2001 Datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/169/136/y2001dat.pdf

      Yeast Drop-Out Bulletin:
      https://www.sigmaaldrich.com/US/en/technical-documents/protocol/microbiological-testing/microbial-culture-media-preparation/yeast-drop-out

      Yeast Growth Protocols page:
      https://www.sigmaaldrich.com/US/en/technical-documents/protocol/microbiological-testing/pathogen-and-spoilage-testing/yeast-growth-protocols

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service