Skip to Content
Merck
All Photos(1)

Key Documents

UC283

Sigma-Aldrich

7-Hydroxycoumarin sulfate potassium salt

≥95% (HPLC)

Synonym(s):

2-Oxo-2H-1-benzopyran-7-yl-sulfate potassium salt, 7-Sulfooxy-2-oxo-2H-1-benzopyran potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5KO6S
Molecular Weight:
280.30
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

solid

color

white to off-white

mp

204-205 °C

solubility

H2O: soluble
methanol: soluble

storage temp.

−70°C

SMILES string

[K+].[O-]S(=O)(=O)Oc1ccc2C=CC(=O)Oc2c1

InChI

1S/C9H6O6S.K/c10-9-4-2-6-1-3-7(5-8(6)14-9)15-16(11,12)13;/h1-5H,(H,11,12,13);/q;+1/p-1

InChI key

SVUZPRLQAASRKC-UHFFFAOYSA-M

Application

Coumarin phase II metabolite.
The product can be used for the assay of coumarin metabolites.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

7-Hydroxycoumarin sulfate potassium salt is soluble in water and methanol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service