Skip to Content
Merck
All Photos(1)

Key Documents

TRUE9

Sigma-Aldrich

TrueGel3D Hydrogel Kits

SLO-PVA, PEG non cell-degradable crosslinker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

The gel is formed by stable thioether bonds between thiol groups on PEG non cell-degradable crosslinker and thio-reactive groups on SLO-PVA. The polymer, SLO-PVA is functionalized with thiol-reactive groups which react with crosslinker at slower rate than maleimides in FAST-PVA. SLO-PVA does not have sites for cell attachment, but it can be customized to conjugate RGD peptide using TrueGel3D RGD peptide (TRUERGD-1EA) prior to the crosslinking step.

Application

TRUE9-1KT is used to generate hydrogels of different stiffness (0.15 to 5 KPa shear modulus) to mimic in-vivo conditions and to investigate the effect of matrix stiffness on cell physiology and behavior.SLO-PVA based gels are used to grow cells that do not need attachment to hydrogel for survival but rather use their secreted extracellular matrix. The SLO-PVA based hydrogels are preferred over dextran-based gels for long-term culture of cells producing carbohydrate degrading enzymes. TrueGel3DTM Hydrogel Kits has been used in the generation of 3D gel based tumor models.

Features and Benefits

  • Slow gelling (takes 6 minutes to 25 minutes, depends on gel stiffness)
  • Flexibility in modulating gel stiffness
  • Does not allow cell migration
  • Does not allow cell recovery and post-culture analyses
  • Cell inert hydrogel (if not customized with RGD peptide)

Components

TRUE9-1KT contains SLO-PVA solution in phosphate buffer and lyophilized PEG non cell-degradable crosslinker with reconstitution buffer and water.

Caution

Avoid prolong exposure of PEG non cell-degradable crosslinker to air and store SLO-PVA in small portions to avoid frequent freeze-thaw cycles.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design of Spherically Structured 3D In vitro Tumor Models-Advances and Prospects
Ferreira LP, et al.
Acta Biomaterialia (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service