Skip to Content
Merck
All Photos(1)

Key Documents

TRUE6

Sigma-Aldrich

TrueGel3D Hydrogel Kits

SLO-DEXTRAN, allow cell recovery, PEG non cell-degradable crosslinker

Synonym(s):

3D Hydrogel Kit, TrueGel3D Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

The gel is formed by stable thioether bonds between thiol groups on PEG non cell-degradable crosslinker and thio-reactive groups on SLO-DEXTRAN. The polymer, SLO-DEXTRAN is functionalized with thiol-reactive groups which react with crosslinker at slower rate than maleimides in FAST-DEXTRAN. SLO-DEXTRAN does not have sites for cell attachment, but it can be customized to conjugate RGD peptide using TrueGel3D RGD peptide (TRUERGD-1EA) prior to the crosslinking step.

TrueGel3D enzymatic cell recovery solution (TRUEENZ-500UL) is used to dissolve dextran polymers for easy recovery of chemically fixed or live cells for post-culture analyses including RT-PCR, western blots and other molecular biological studies.

Application

TRUE6-1KT is used to generate hydrogels of different stiffness (0.15 to 5 KPa shear modulus) to mimic natural tissue stiffness and to investigate the effect of mechanical stimulus on cell physiology and behavior. It is also used to culture non-proliferating cells and proliferating spheroids that do not attach to hydrogel for survival, but rather use their own extracellular matrix.

Features and Benefits

  • Slow gelling (takes 8 minutes to 25 minutes, depends on gel stiffness)
  • Flexibility in modulating gel stiffness
  • Does not allow cell migration
  • Allow cell recovery and post-culture analyses
  • Cell inert hydrogel (if not customized with RGD peptide)

Components

TRUE6-1KT contains SLO-DEXTRAN solution in phosphate buffer and lyophilized PEG non cell-degradable crosslinker with reconstitution buffer and water.

Caution

Avoid prolong exposure of PEG non cell-degradable crosslinker to air.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service