Skip to Content
Merck
All Photos(2)

Key Documents

T6069

Sigma-Aldrich

Thio-TEPA

Synonym(s):

N,N′,N″-Triethylenethiophosphoramide, Thio-Tep, Thiofozil, Thiotef, Tio-tef, Tiofosfamid, Tiofosyl, Tiofozil, Tris(aziridinyl)phosphine sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N3PS
CAS Number:
Molecular Weight:
189.22
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

form

solid

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

color

white

antibiotic activity spectrum

neoplastics

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

S=P(N1CC1)(N2CC2)N3CC3

InChI

1S/C6H12N3PS/c11-10(7-1-2-7,8-3-4-8)9-5-6-9/h1-6H2

InChI key

FOCVUCIESVLUNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aziridine

Application

A polyfunctional alkylating agent

Biochem/physiol Actions

The unstable nitrogen-carbon groups alkylate with DNA which causes irreversible DNA damage. They stop tumor growth by crosslinking guanine nucleobases in DNA double-helix strands, directly attacking DNA. The DNA strands are unable to uncoil and separate which halts cell division.

Analysis Note

Solubility - freely soluble in water , in chloroform and in ethanol (96%)

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Carc. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel A Bobholz et al.
Tomography (Ann Arbor, Mich.), 6(2), 160-169 (2020-06-18)
Magnetic resonance (MR)-derived radiomic features have shown substantial predictive utility in modeling different prognostic factors of glioblastoma and other brain cancers. However, the biological relationship underpinning these predictive models has been largely unstudied, and the generalizability of these models had
Ki Woong Sung et al.
Neuro-oncology, 15(3), 352-359 (2012-12-22)
We assessed the feasibility and effectiveness of reduced-dose craniospinal (CS) radiotherapy (RT) followed by tandem high-dose chemotherapy and autologous stem cell transplantation (HDCT/autoSCT) in reducing late adverse effects without jeopardizing survival among children with high-risk medulloblastoma (MB). From October 2005
B Kasenda et al.
Annals of oncology : official journal of the European Society for Medical Oncology, 23(10), 2670-2675 (2012-04-05)
High-dose chemotherapy followed by autologous stem-cell transplantation (HCT-ASCT) is a promising approach in eligible patients with primary central nervous system lymphoma (PCNSL). We report long-term data of patients who were treated according to HCT-ASCT containing protocols. We analyzed survival and
Michael P Fay et al.
Statistics in medicine, 31(28), 3760-3772 (2012-07-13)
We consider weighted logrank tests for interval censored data when assessment times may depend on treatment, and for each individual, we only use the two assessment times that bracket the event of interest. It is known that treating finite right
Soo Hyeon Bae et al.
Chemico-biological interactions, 205(1), 11-19 (2013-06-20)
The present study was performed to evaluate the potency and specificity of sibutramine as an inhibitor of the activities of nine human CYP isoforms in liver microsomes. Using a cocktail assay, the effects of sibutramine on specific marker reactions of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service