Skip to Content
Merck
All Photos(1)

Key Documents

T1287

Sigma-Aldrich

Tetramethylrhodamine isothiocyanate–Dextran

average mol wt 155,000

Synonym(s):

TRITC-Dextran

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

synthetic (organic)

form

powder

mol wt

average mol wt 155,000

extent of labeling

~1 TRITC per 100 glucose residues

color

purple to pink

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Tetramethylrhodamine isothiocyanate-Dextran/TRITC-Dextran is a fluorochrome.

Application

Tetramethylrhodamine isothiocyanate-Dextran has been used:
  • as fluorescent dyes
  • to test a range of molecule sizes in dextran transport assays
  • to label blood plasma in order to visualize the vasculature

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hematopoietic Cancers
Immune Response, 1025-1063 (2006)
Wendy Leskova et al.
Investigative ophthalmology & visual science, 61(10), 12-12 (2020-08-08)
Although it is known that the retinal arteriolar vasculature is constricted in hypertension, the details of retinal hemodynamics and perfusion of the retinal circulation have yet to be adequately characterized. Male and female spontaneously hypertensive rats (SHR) and Wistar-Kyoto (WKY)
David Entenberg et al.
Methods (San Diego, Calif.), 128, 65-77 (2017-09-16)
Pathologists rely on microscopy to diagnose disease states in tissues and organs. They utilize both high-resolution, high-magnification images to interpret the staining and morphology of individual cells, as well as low-magnification overviews to give context and location to these cells.
Monika I Linder et al.
Developmental cell, 43(2), 141-156 (2017-10-25)
During interphase, the nuclear envelope (NE) serves as a selective barrier between cytosol and nucleoplasm. When vertebrate cells enter mitosis, the NE is dismantled in the process of nuclear envelope breakdown (NEBD). Disassembly of nuclear pore complexes (NPCs) is a
The formation of a functional retinal pigment epithelium occurs on porous polytetrafluoroethylene substrates independently of the surface chemistry
Kearns V R, et al.
Journal of Materials Science. Materials in Medicine, 28(8), 124-124 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service