Skip to Content
Merck
All Photos(1)

Key Documents

SRP4702

Sigma-Aldrich

CTGF human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

connective tissue growth factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

501.0-2.0 μg/mL ED50

mol wt

~11.2 kDa

packaging

pkg of 20 μg

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CTGF(1490)

General description

The gene CTGF (connective tissue growth factor) is mapped to human chromosome 6q23.2. CTGF is a member of the CCN (Cyr61, CTGF , Nov) family of secreted cysteine rich regulatory proteins and is the major mitogenic and chemoattractant protein produced by umbilical vein and vascular endothelial cells. The protein has an N-terminal secretory signal peptide and four domains (an insulin-like growth factor-binding protein domain, a Von Willebrand factor domain, a thrombospondin type-1 repeat module, and a CT). It is expressed in various tissues, such as connective tissue, heart, brain, kidney, lung, liver, muscle, pancreas gland and placenta. Recombinant human CTGF is a 11.2Da protein of 98 amino acid residues.

Biochem/physiol Actions

CTGF (connective tissue growth factor) stimulates the proliferation and differentiation of chondrocytes, induces angiogenesis, promotes cell adhesion of fibroblasts, endothelial, and epithelial cells. It binds to IGF (insulin growth factor), TGF (transforming growth factor) β1 and BMP (bone morphogenetic protein)-4. CTGF plays a crucial role in the progression of diabetic nephropathy. It can work as an oncoprotein in glioma and melanoma. However, it works as a tumor suppressor in lung cancer and colon cancer.

Physical form

Sterile filtered and Lyophilized without additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. The solution can then be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hsiao-Chi Tsai et al.
Oncotarget, 5(11), 3800-3812 (2014-07-09)
Osteosarcoma, the most common primary malignant bone tumor, shows potent capacity for local invasion and distant metastasis. Connective tissue growth factor (CTGF/CCN2), a secreted protein, binds to integrins, modulates invasive behavior of certain human cancer cells. Effect of CTGF in
Promoter hypermethylation contributes to frequent inactivation of a putative conditional tumor suppressor gene connective tissue growth factor in ovarian cancer.
Kikuchi R, et al.
Cancer Research, 67, 7095-7105 (2007)
Epithelial derived CTGF promotes breast tumor progression via inducing EMT and collagen I fibers deposition.
Zhu X, et al.
Oncotarget, 6, 25320-25338 (2015)
Emerging role of CCN family proteins in tumorigenesis and cancer metastasis (Review).
Li J, et al.
International Journal of Molecular Medicine, 36, 1451-1463 (2015)
Potential Renoprotective Agents through Inhibiting CTGF/CCN2 in Diabetic Nephropathy.
Wang S, et al.
Journal of Diabetes Research, 2015, 962383-962383 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service