Skip to Content
Merck
All Photos(3)

Key Documents

SRP3186

Sigma-Aldrich

Human Vitronectin

recombinant, expressed in HEK 293 cells, powder, suitable for cell culture

Synonym(s):

Epiboin, S-Protein, Serum-spreading factor, V75, VTN

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

Product Name

Vitronectin human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

≤0.5 μg/mL EC50

mol wt

52.2 kDa (calculated)

packaging

pkg of 250 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

solubility

water: soluble

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VTN(7448)

General description

Vitronectin is a secreted glycoprotein which is synthesized in the liver. It is also secreted by respiratory epithelial cells and is present in healthy lungs. It circulates primarily in monomeric form, but can undergo conformational change to a structure that forms disulfide linked multimers.The vitronectin gene consists of approximately 4.5–5 kb base pairs with eight exons and seven introns.
Recombinant human Vitronectin is a protein consisting of 459 amino acids. It exists in two forms: a single chain and a clipped form consisting of two chains held together by disulfide bonds resulting from an endogenous cleavage. Recombinant vitronectin can function as a chemically defined matrix component in human embryonic stem cell renewal media.

Application

Vitronectin human has been used to study the effects of ECM (extracellular matrix) ligands on C6 astroglioma cells. It has also been used as an attachment-promoting protein for coating the cell plates.

Biochem/physiol Actions

The multimeric vitronectin protein can efficiently bind to and incorporate into the extracellular matrix. Within the matrix, Vitronectin can support cell adhesion through binding to various integrins and other proteoglycans. Its interaction with integrin αvβ3 enhances hypoxia resistance and tumor cell invasion. It is also associated with cancer progression in various cancers. It is also involved in wound repair and controls complement system as wel. Moreover, vitronectin functions as a marker of profibrotic activity in liver, heart and kidney.

Features and Benefits

  • Human-derived vitronectin
  • Produced through recombinant expression in HEK 293 cells
  • Low endotoxin levels

Sequence

DQESCKGRCT EGFNVDKKCQ CDELCSYYQS CCTDYTAECK PQVTRGDVFT MPEDEYTVYD DGEEKNNATV HEQVGGPSLT SDLQAQSKGN PEQTPVLKPE EEAPAPEVGA SKPEGIDSRP ETLHPGRPQP PAEEELCSGK PFDAFTDLKN GSLFAFRGQY CYELDEKAVR PGYPKLIRDV WGIEGPIDAA FTRINCQGKT YLFKGSQYWR FEDGVLDPDY PRNISDGFDG IPDNVDAALA LPAHSYSGRE RVYFFKGKQY WEYQFQHQPS QEECEGSSLS AVFEHFAMMQ RDSWEDIFEL LFWGRTSAGT RQPQFISRDW HGVPGQVDAA MAGRIYISGM APRPSLAKKQ RFRHRNRKGY RSQRGHSRGR NQNSRRPSRA TWLSLFSSEE SNLGANNYDD YRMDWLVPAT CEPIQSVFFF SGDKYYRVNL RTRRVDTVDP PYPRSIAQYW LGCPAPGHL

Physical form

Lyophilized from Phosphate Buffered Saline (PBS), pH 7.4, plus 2.5% trehalose.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Other Notes

Under reducing conditions, the single chain migrates at an apparent molecular mass of 75kDa on SDS-PAGE, and the clipped form migrates at molecular masses of 65kDa and 10kDa.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extracellular matrices and cell adhesion.
E Ruoslahti et al.
Arteriosclerosis (Dallas, Tex.), 5(6), 581-594 (1985-11-01)
Vitronectin expression in the airways of subjects with asthma and chronic obstructive pulmonary disease.
Salazar-Pelaez LM, et al.
PLoS ONE, 10, e0119717-e0119717 (2015)
Stefan R Braam et al.
Stem cells (Dayton, Ohio), 26(9), 2257-2265 (2008-07-05)
Defined growth conditions are essential for many applications of human embryonic stem cells (hESC). Most defined media are presently used in combination with Matrigel, a partially defined extracellular matrix (ECM) extract from mouse sarcoma. Here, we defined ECM requirements of
Distribution of vitronectin mRNA during murine development.
Seiffert D, et al.
Developmental Dynamics, 203, 71-79 (1995)
Denise Salzig et al.
Stem cells international, 2016, 5246584-5246584 (2016-03-24)
The manufacture of human mesenchymal stem cells (hMSCs) for clinical applications requires an appropriate growth surface and an optimized, preferably chemically defined medium (CDM) for expansion. We investigated a new protein/peptide-free CDM that supports the adhesion, growth, and detachment of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service