V0132
Rat Vitronectin
from rat plasma, powder, suitable for cell culture
Synonym(s):
Serum spreading factor
About This Item
Recommended Products
product name
Vitronectin from rat plasma, lyophilized powder, BioReagent, suitable for cell culture
biological source
rat plasma
Quality Level
product line
BioReagent
form
lyophilized powder
mol wt
75 kDa
packaging
pkg of 50 μg
technique(s)
cell culture | mammalian: suitable
surface coverage
0.1 μg/cm2
solubility
water: soluble 1.00 mL, clear, colorless
UniProt accession no.
shipped in
dry ice
storage temp.
2-8°C
Gene Information
rat ... Vtn(29169)
General description
Application
Biochem/physiol Actions
Components
Caution
Preparation Note
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service