Skip to Content
Merck
All Photos(1)

Key Documents

SRP3143

Sigma-Aldrich

Pleiotrophin human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Heparin Affin Regulatory Protein (HARP), Heparin-binding growth factor-8(HBGF-8), Osteoblast-Specific Factor-1 (OSF-1), PTN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

15.4 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PTN(5764)

General description

PTN (pleiotrophin) is a 136-residue, platelet-derived growth factor (PDGF)-inducible, heparin-binding cytokine. It shares ~50% similarity with midkine, a retinoic acid-inducible factor. It is expressed in a tissue and stage dependent manner during development, but in adults, this protein is expressed constitutively, though in a restricted tissue pattern. PTN is a low molecular weight protein composed of two domains linked by disulfide bridges, with three antiparallel β-sheets in each domain. This gene is localized to human chromosome 7q33-q34. Recombinant human Pleiotrophin is a 15.4 kDa protein containing 136 amino acid residues and five intra-molecular disulfide bonds.

Biochem/physiol Actions

PTN (pleiotrophin) is a ligand for receptor protein tyrosine phosphatase (RPTP) β/ζ, and binds with it and inhibits its catalytic capabilities in a “ligand-dependent receptor inactivation”-mechanism, and promotes tyrosine phosphorylation of β-catenin. It acts as a mitogen for fibroblasts, endothelial cells, epithelial cells, and trophoblasts. It promotes neurite outgrowth and glial process outgrowth from neurons and glial progenitor cells, respectively. This protein functions as a protooncogene, and is up-regulated in multiple types of tumors, where it promotes tumor angiogenesis and progression. This protein functions as a crucial mitogen during nerve, bone, vascular, and tumor growth, and has been found to be up-regulated in macrodactyly, identifying it as a candidate gene for macrodactyly.

Sequence

GKKEKPEKKV KKSDCGEWQW SVCVPTSGDC GLGTREGTRT GAECKQTMKT QRCKIPCNWK KQFGAECKYQ FQAWGECDLN TALKTRTGSL KRALHNAECQ KTVTISKPCG KLTKPKPQAE SKKKKKEGKK QEKMLD

Physical form

Lyophilized from 10 mM sodium phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pleiotrophin signals increased tyrosine phosphorylation of beta beta-catenin through inactivation of the intrinsic catalytic activity of the receptor-type protein tyrosine phosphatase beta/zeta.
Meng K
Proceedings of the National Academy of Sciences of the USA, 97, 2603-2608 (2000)
Midkine and pleiotrophin in neural development and cancer
Kadomatsu K and Muramatsu T
Cancer Letters, 204, 127-143 (2004)
Frank H Lau et al.
PloS one, 7(7), e40423-e40423 (2012-08-01)
Macrodactyly is a rare family of congenital disorders characterized by the diffuse enlargement of 1 or more digits. Multiple tissue types within the affected digits are involved, but skeletal patterning and gross morphological features are preserved. Not all tissues are
Takashi Muramatsu
Journal of biochemistry, 132(3), 359-371 (2002-09-03)
Midkine (MK) and pleiotrophin (PTN) are low molecular weight proteins with closely related structures. They are mainly composed of two domains held by disulfide bridges, and there are three antiparallel beta-sheets in each domain. MK and PTN promote the growth
T Muramatsu
The International journal of developmental biology, 37(1), 183-188 (1993-03-01)
Using mouse teratocarcinoma system, we found a novel retinoic acid responsive gene. Midkine (MK), the product of the gene is a secreted, heparin-binding protein of molecular weight 14,000. MK gene is intensely expressed in the midgestation period, and in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service