Skip to Content
Merck
All Photos(1)

Key Documents

SRP3111

Sigma-Aldrich

MDC (67 aa) (CCL22) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

ABCD-1, CCL22, Macrophage-Derived Chemokine, STCP-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

8.0 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL22(6367)

General description

Macrophage-derived chemokine (MDC) is also known as C-C motif chemokine ligand 22 (CCL22). It is a CC chemokine that is produced in B cells, macrophages, monocyte-derived dendritic cells, activated natural killer (NK) cells and cluster of differentiation 4 (CD4) T cells. The gene encoding it is localized on human chromosome 16. Recombinant human MDC is an 8.0kDa protein containing 67 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.

Biochem/physiol Actions

Macrophage-derived chemokine (MDC) signals through the C-C chemokine receptor type 4 (CCR4). It chemo-attracts monocytes, dendritic cells and natural killer (NK) cells and aids in their function. The protein exerts human immunodeficiency virus (HIV) suppressive activity. The 67 amino acid form of MDC displays reduced chemoattractant activity but retains HIV suppressive activity. It is expressed in various cancers and is involved in recruitment of regulatory T cells (Treg) within a tumor.

Sequence

YGANMEDSVC CRDYVRYRLP LRVVKHFYWT SDSCPRPGVV LLTFRDKEIC ADPRVPWVKM ILNKLSQ

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Fan Leung et al.
Pediatric allergy and immunology : official publication of the European Society of Pediatric Allergy and Immunology, 14(4), 296-301 (2003-08-13)
Chemokines are responsible for the trafficking of leukocytes to sites of inflammation. Serum chemokine levels were previously shown to be increased in adult patients with atopic dermatitis (AD). We tested whether serum concentrations of chemokines, including macrophage-derived chemokine (MDC), thymus
S Struyf et al.
Journal of immunology (Baltimore, Md. : 1950), 161(6), 2672-2675 (1998-09-22)
Posttranslational processing of chemokines increases (IL-8) or decreases (monocyte chemotactic protein-1) their chemotactic potency. Macrophage-derived chemokine (MDC) attracts monocytes, dendritic cells, activated lymphocytes, and NK cells and has reportedly anti-HIV-1 activity. Here we report that truncation of MDC by deletion
Soluble factors from T cells inhibiting X4 strains of HIV are a mixture of ? chemokines and RNases.
Cocchi F
Proceedings of the National Academy of Sciences of the USA, 109(14), 5411-5416 (2012)
Genetic Polymorphisms of CCL22 and CCR4 in Patients with Lung Cancer.
Erfani N
Iranian Journal of Medical Sciences, 39(4), 367-373 (2014)
Human macrophage-derived chemokine (MDC), a novel chemoattractant for monocytes, monocyte-derived dendritic cells, and natural killer cells.
Godiska R
The Journal of Experimental Medicine, 185(9), 1595-1604 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service