SRE0038
β-Glucuronidase from abalone
lyophilized powder, β-glucuronidase ≥1,500,000 units/g solid
About This Item
Recommended Products
biological source
abalone
form
lyophilized powder
β-glucuronidase activity
≥1,500,000 units/g solid
secondary activity
≤300,000 units/g solid sulfatase
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
Learn more
about recent application data generated by Sigma R&D to optimize hydrolysis for different drug classes using enzymes from different sources and the use of a chromatographicaly purified enzyme to reduce the effect of esterase activity resulting in conversion of 6-MAM to Morphine.
Unit Definition
Physical form
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1 - Skin Sens. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service