Skip to Content
Merck
All Photos(1)

Key Documents

F6020

Sigma-Aldrich

Fenofibrate

≥99% (TLC), powder, PPARα agonist

Synonym(s):

2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic acid isopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21ClO4
CAS Number:
Molecular Weight:
360.83
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Fenofibrate, ≥99%, powder

Assay

≥99%

form

powder

color

off-white

originator

Abbott

storage temp.

room temp

SMILES string

CC(C)OC(=O)C(C)(C)Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2

InChI

1S/C20H21ClO4/c1-13(2)24-19(23)20(3,4)25-17-11-7-15(8-12-17)18(22)14-5-9-16(21)10-6-14/h5-13H,1-4H3

InChI key

YMTINGFKWWXKFG-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Fenofibrate has been used:
  • to study its effect on plasma lipids, liver phenotype and gene expression
  • to study its impact on endothelium-dependent vasodilatation of thoracic aorta
  • to administer to NASH knock-in mice along with a high fat diet (HFD) to study its effect

Biochem/physiol Actions

Fenofibrate increases high density lipoprotein levels by reducing cholesteryl ester transfer protein expression. It is used in treating the condition of increased cholesterol levels in the blood, abnormal lipid levels in the body and also hypertriglyceridaemia. Fenofibrate is a lipid regulating drug and proliferator-activated receptor-α (PPARα) mediates its action. It decreases the plasma levels of fibrinogen and C-reactive protein. By this fenofibrate allows better flow-mediated dilatation and reduces the risk of atherosclerosis. Fenofibrate is also known to reduce uric acid levels.

Features and Benefits

This compound is featured on the Nuclear Receptors (PPARs) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Oral

Target Organs

Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Early diet-induced non-alcoholic steatohepatitis in APOE2 knock-in mice and its prevention by fibrates
Shiri-Sverdlov R, et al.
Journal of Hepatology, 44(4), 732-741 (2006)
Fenofibrate
Keating GM and Croom KF
Drugs, 67(1), 121?153-121?153 (2012)
Intrahepatic cholesterol influences progression, inhibition and reversal of non-alcoholic steatohepatitis in hyperlipidemic mice
Wouters K, et al.
Febs Letters, 584(5), 1001-1005 (2010)
Oshri Avraham et al.
eLife, 10 (2021-09-30)
Sensory neurons with cell bodies in dorsal root ganglia (DRG) represent a useful model to study axon regeneration. Whereas regeneration and functional recovery occurs after peripheral nerve injury, spinal cord injury or dorsal root injury is not followed by regenerative
Sujeong Park et al.
Experimental & molecular medicine, 53(7), 1159-1169 (2021-07-22)
In this study, we hypothesized that deregulation in the maintenance of the pool of coenzyme A (CoA) may play a crucial role in the pathogenesis of nonalcoholic fatty liver disease (NAFLD). Specific deletion of Acot12 (Acot12-/-), the major acyl-CoA thioesterase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service