Skip to Content
Merck
All Photos(1)

Key Documents

SML2117

Sigma-Aldrich

TCS-1102

≥98% (HPLC)

Synonym(s):

BBAC, (2S)-N-[1,1′-Biphenyl]-2-yl-1-[2-[(1-methyl-1H-benzimidazol-2-yl)thio]acetyl]-2-pyrrolidinecarboxamide, DORA-1, N-(1,1′-Biphenyl-2-yl)-1-{[(1-methyl-1H-benzimidazol-2-yl)thio]acetyl}-L-prolinamide, TCS 1102, TCS1102, (2S)-N-[1,1′-Biphenyl]-2-yl-1-[2-[(1-methyl-1H-benzimidazol-2-yl)thio]acetyl]-2-pyrrolidinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H26N4O2S
CAS Number:
Molecular Weight:
470.59
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN1C2=CC=CC=C2N=C1SCC(N3CCC[C@H]3C(NC4=CC=CC=C4C5=CC=CC=C5)=O)=O

InChI

1S/C27H26N4O2S/c1-30-23-15-8-7-14-22(23)29-27(30)34-18-25(32)31-17-9-16-24(31)26(33)28-21-13-6-5-12-20(21)19-10-3-2-4-11-19/h2-8,10-15,24H,9,16-18H2,1H3,(H,28,33)/t24-/m0/s1

InChI key

YSBGRVXJEMSEQY-DEOSSOPVSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

TCS-1102 (DORA-1) is a high-affinity (Ki = 3 nM against [3H]-TCS-1102 for binding hOX1R and Ki = 0.2 nM against [3H]-pyrrolylphenyl analog for binding hOX2R) dual orexin receptor antagonist (DORA) that potently inhibits calcium mobilization in hOX1R & hOX2R CHO transfectants (IC50 = 17 & 4 nM, respectively). TCS-1102 promotes sleep (50-100 mg/kg p.o.), decreases fear/anxiety (10-20 mg/kg i.p.), and inhibits ADL-orexin B peptide-induced locomotion (by 56%/74%/100% with 15/50/100 mg/kg i.p. 30 min before ADL-XB ICV injection) with good pharmacokinetic properties, brain penetration (Brain/plasma conc. = 2.37 μM/3.50 μM 3 hrs post 100 mg/kg i.p.), and oral availability (Cmax/AUC(0–24h) = 1.21 μM/6.8 μM h and 3.01 μM/12.5 μM h, respectively post 50 and 100 mg/kg p.o. dosage) in rats in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vijayakumar Mavanji et al.
Sleep, 38(9), 1361-1370 (2015-04-08)
The ventrolateral preoptic area (VLPO) and the orexin/hypocretin neuronal system are key regulators of sleep onset, transitions between vigilance states, and energy homeostasis. Reciprocal projections exist between the VLPO and orexin/hypocretin neurons. Although the importance of the VLPO to sleep
Huiying Wang et al.
Behavioural brain research, 316, 47-53 (2016-08-31)
Orexin (hypocretin) neurons located in the posterior hypothalamus send projections to multiple areas of the brain involved in arousal and experimental evidence indicates that these neurons play a role in the physiological and behavioral responses to stress. This study was
Yuhei Nishimura et al.
Frontiers in pharmacology, 6, 257-257 (2015-11-19)
Sleep-wake states are impaired in various neurological disorders. Impairment of sleep-wake states can be an early condition that exacerbates these disorders. Therefore, treating sleep-wake dysfunction may prevent or slow the development of these diseases. Although many gene products are likely
Jeffrey M Bergman et al.
Bioorganic & medicinal chemistry letters, 18(4), 1425-1430 (2008-01-22)
A series of OX(2)R/OX(1)R dual orexin antagonists was prepared based on a proline bis-amide identified as a screening lead. Through a combination of classical and library synthesis, potency enhancing replacements for both amide portions were discovered. N-methylation of the benzimidazole
Xiaoyu Chen et al.
Brain structure & function, 219(6), 2103-2118 (2013-08-21)
Orexins (hypocretins) are peptides that have been shown to regulate behavioral arousal and wakefulness. Recent evidence indicates that orexin neurons are activated by stress and that orexins play a role in anxiety. The present paper describes a series of experiments

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service