Skip to Content
Merck
All Photos(1)

Key Documents

S2694

Sigma-Aldrich

SB-408124

≥98% (HPLC), solid

Synonym(s):

N-(6,8-Difluoro-2-methyl-4-quinolinyl)-N′-[4-(dimethylamino)phenyl]urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18F2N4O
CAS Number:
Molecular Weight:
356.37
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: >10 mg/mL

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

CN(C)c1ccc(NC(=O)Nc2cc(C)nc3c(F)cc(F)cc23)cc1

InChI

1S/C19H18F2N4O/c1-11-8-17(15-9-12(20)10-16(21)18(15)22-11)24-19(26)23-13-4-6-14(7-5-13)25(2)3/h4-10H,1-3H3,(H2,22,23,24,26)

InChI key

JTARFZSNUAGHRB-UHFFFAOYSA-N

Application

SB-408124 was used to block orexin A-mediated cardiovascular effects in rats. It was used to study block the increase of endogenous glucose production mediated by bicuculline in rats.

Biochem/physiol Actions

SB 408124 is an OX1 orexin receptor antagonist.
SB-408124 blocks the orexin type 1 receptor that stimulates the production of stress hormone in hypothalamus. It blocks orexin A-induced increase in arterial pressure and elevation of heart rate in rats. SB-408124 modulates phrenic nerve activity that is important in respiration.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Willis K Samson et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 292(1), R382-R387 (2006-08-12)
Hypocretin/orexin acts pharmacologically in the hypothalamus to stimulate stress hormone secretion at least in part by an action in the hypothalamic paraventricular nucleus, where the peptide's receptors have been localized. In addition, orexin acts in the brain to increase sympathetic
Andrea Corcoran et al.
Advances in experimental medicine and biology, 669, 109-113 (2010-03-11)
Release of hypocretins (orexins) by neurons in the lateral hypothalamus is an important contributor to arousal state, thermoregulation, feeding behavior, and has recently been proposed to play a role in breathing and central chemosensitivity. Using the in situ arterially perfused
Kaiyu Xu et al.
Nature communications, 11(1), 3594-3594 (2020-07-19)
Circular RNAs (circRNAs) are abundant in mammalian brain and some show age-dependent expression patterns. Here, we report that circGRIA1, a conserved circRNA isoform derived from the genomic loci of α-mino-3-hydroxy-5-methyl-4-isoxazole propionic acid (AMPA) receptor subunit Gria1, shows an age-related and
Chun-Xia Yi et al.
Diabetes, 58(9), 1998-2005 (2009-07-14)
The hypothalamic neuropeptide orexin influences (feeding) behavior as well as energy metabolism. Administration of exogenous orexin-A into the brain has been shown to increase both food intake and blood glucose levels. In the present study, we investigated the role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service