Skip to Content
Merck
All Photos(1)

Key Documents

SML2001

Sigma-Aldrich

GSK′843

≥98% (HPLC)

Synonym(s):

3-(5-Benzothiazolyl)-7-(1,3-dimethyl-1H-pyrazol-5-yl)-thieno[3,2-c]pyridin-4-amine, GSK 843, GSK-843, GSK843

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15N5S2
CAS Number:
Molecular Weight:
377.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c2c(nc1)cc(cc2)c3c4c([s]c3)c(cnc4N)c5[n](nc(c5)C)C

InChI key

BPKSNNJTKPIZKR-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

A potent ATP-competitive RIP3 (RIPK3) kinase inhibitor that suppresses necrosis induction in both human and murine cell cultures.
GSK′843 is a potent ATP-competitive RIP3 (RIPK3) kinase inhibitor (IC50 = 6.5 nM) with much reduced or little affinity toward 262 other kinases (IC50 >1 μM against 10 μM ATP in competitive binding assay), including RIP5. GSK′843 effectively suppresses TNF/Z-VAD-FMK/IAP agonist treatment-induced necrosis induction in both human and murine cell cultures (EC50 ranges from <0.12 to 3 μM), while inducing apoptosis in the absence of caspase inhibitor as a result of caspase 8 activation via RHIM-driven recruitment to assemble a Casp8-FADD-cFLIP complex independent of pronecrotic kinase activities and MLKL.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pratyusha Mandal et al.
Molecular cell, 56(4), 481-495 (2014-12-03)
Receptor-interacting protein kinase 3 (RIP3 or RIPK3) has emerged as a central player in necroptosis and a potential target to control inflammatory disease. Here, three selective small-molecule compounds are shown to inhibit RIP3 kinase-dependent necroptosis, although their therapeutic value is
William J Kaiser et al.
The Journal of biological chemistry, 288(43), 31268-31279 (2013-09-11)
Toll-like receptor (TLR) signaling is triggered by pathogen-associated molecular patterns that mediate well established cytokine-driven pathways, activating NF-κB together with IRF3/IRF7. In addition, TLR3 drives caspase 8-regulated programmed cell death pathways reminiscent of TNF family death receptor signaling. We find
N Preyat et al.
Cell death and differentiation, 23(1), 29-40 (2015-05-23)
Cellular necrosis has long been regarded as an incidental and uncontrolled form of cell death. However, a regulated form of cell death termed necroptosis has been identified recently. Necroptosis can be induced by extracellular cytokines, pathogens and several pharmacological compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service